Skip to Content
Merck
All Photos(2)

Key Documents

5.04744

Sigma-Aldrich

Aphidicolin

≥98% (TLC), liquid, DNA polymerase α and δ inhibitor, Calbiochem

Synonym(s):

InSolution Aphidicolin

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€186.00

€186.00


Estimated to ship on24 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€186.00

About This Item

Empirical Formula (Hill Notation):
C20H34O4
CAS Number:
Molecular Weight:
338.48
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€186.00


Estimated to ship on24 May 2025


Request a Bulk Order

Product Name

Aphidicolin, InSolution, ≥98%, 30 mM in DMSO, A cell-permeable tetracyclic diterpene antibiotic

Quality Level

Assay

≥98% (TLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles
desiccated (hygroscopic)
protect from light

storage temp.

−70°C

SMILES string

O[C@]1([C@H]2C[C@@]3([C@@]4([C@H]([C@@]([C@@H](CC4)O)(CO)C)CC[C@H]3C2)C)CC1)CO

InChI

1S/C20H34O4/c1-17(11-21)15-4-3-13-9-14-10-19(13,7-8-20(14,24)12-22)18(15,2)6-5-16(17)23/h13-16,21-24H,3-12H2,1-2H3/t13-,14+,15-,16+,17-,18-,19-,20-/m0/s1

InChI key

NOFOAYPPHIUXJR-APNQCZIXSA-N

General description

A cell-permeable tetracyclic diterpene antibiotic. Cell synchronization agent. Blocks the cell cycle at the early S-phase. Specific, non-competitive and reversible inhibitor of DNA polymerase α and δ in eukaryotic cells and in some viruses of animal origin. Potentiates apoptosis induced by arabinosyl nucleosides in leukemia cell lines. Also induces apoptosis in HeLaS3 cells, but inhibits vincristine (>Cat. No. 677181)-induced apoptosis in the p53-negative human prostate cancer cell line PC-3.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
DNA polymerase α and δ
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

A 30 mM (1 mg/98 µL) sterile-filtered solution of Aphidicolin (Cat. No. 178273) in DMSO.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C). Aliquots are stable for up to 6 months at -70°C. Use only fresh DMSO for further dilutions.

Other Notes

Borner, M.M., et al. 1995. Cancer Res.55, 2122.
Poluha, W., et al. 1995. Oncogene10, 185.
Urbani, L., et al. 1995. Exp. Cell Res.219, 159.Schimke, R.T., et al. 1994. Philos. Trans. R. Soc. London. B. Biol. Sci.345, 311.Kuwakado, K., et al. 1993. Biochem. Pharmacol.46, 1909.Hubermann, J.A. 1981. Cell23, 647.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - (refers to pure substance)

Flash Point(C)

87 °C - (refers to pure substance)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service