Skip to Content
Merck
All Photos(1)

Key Documents

5.04537

Sigma-Aldrich

PKM2 Activator III

Synonym(s):

PKM2 Activator III, Pyruvate Kinase M2 Activator III, N-(4-(4-(2-Methoxyphenyl)piperazine-1-carbonyl)phenyl)quinoline-8-sulfonamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€265.00

€265.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€265.00

About This Item

Empirical Formula (Hill Notation):
C27H26N4O4S
CAS Number:
Molecular Weight:
502.58
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€265.00


Estimated to ship on28 May 2025


Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 5 mg/mL

storage temp.

2-8°C

General description

A cell-permeable quinoline-sulfonamide that acts as a potent allosteric PKM2 activator both in cell-free enzymatic assays (AC50 = 17 nM with 40 ng recombinant PKM2/200 µL) and in cultures (AC50 = 45 nM in A549 cells) via a high affinity (no dissociation in >1.5 h), 2:1 (compound to PKM2 tetramer) stoichiometric binding, effectively locking PKM2 in an active tetrameric state that is resistant to known intracellular negative regulators of PKM2 tetramer induced by the natural activator FBP (fructose 1,6-bisphosphate). PKM2 stimulation by compound treatment is shown to result in decreased serine biosynthesis (by 56%; 500 nM for 24 h) with concomitant increase in serine influx as a compensating mechanism for maintaining cellular serine level necessary for supporting A549 proliferation. Simultaneous PKM2 activation by drug treatment and culture serine withdrawal results in depletion of cellular serine pool (by ~70% in 24 h) and induction of cytostatic A549 growth arrest (by 56%; 72 h 1 µM drug treatment in BME + NEAA - Ser), but not apoptosis. Serine-dependent proliferation inhibition upon PKM2 activation is reported to be cell-specific, while it is observed in A549 and H460 cultures, SW480 and H522 are not affected.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKM2
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kung, C., et al. 2012. Chem. Biol.19, 1187.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service