Skip to Content
Merck
All Photos(1)

Key Documents

420205

Sigma-Aldrich

Histone Lysine Demethylase Inhibitor VIII, GSK-J4

Histone Lysine Demethylase Inhibitor VIII, GSK-J4, CAS 1373423-53-0, is converted to GSK-J1 intracellularly for effective JMJD3 and UTX inhibition (~ 25 to 50 µM in HeLa cultures).

Synonym(s):

Histone Lysine Demethylase Inhibitor VIII, GSK-J4, GSK-J1 Pro-Drug, JHDM Inhibitor II Pro-Drug, Ethyl-3-(6-(4,5-dihydro-1H-benzo[d]azepin-3(2H)-yl)-2-(pyridin-2-yl)pyrimidin-4-ylamino)propanoate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€250.00

€250.00


Estimated to ship on30 March 2025



Select a Size

Change View
10 MG
€250.00

About This Item

Empirical Formula (Hill Notation):
C24H27N5O2
CAS Number:
Molecular Weight:
417.50
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€250.00


Estimated to ship on30 March 2025


Quality Level

Assay

≥98% (HPLC)

form

semisolid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

tan

solubility

DMSO: 100 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

N3(CCc4c(cccc4)CC3)c1nc(nc(c1)NCCC(=O)OCC)c2ncccc2

InChI

1S/C24H27N5O2/c1-2-31-23(30)10-14-26-21-17-22(28-24(27-21)20-9-5-6-13-25-20)29-15-11-18-7-3-4-8-19(18)12-16-29/h3-9,13,17H,2,10-12,14-16H2,1H3,(H,26,27,28)

InChI key

WBKCKEHGXNWYMO-UHFFFAOYSA-N

General description

A cell-permeable GSK-J1 (Cat. No. 420204) prodrug that is ineffective in cell-free assays, but is effectively converted to GSK-J1 intracellularly for effective JMJD3 and UTX inhibition (effective conc. 25 to 50 µM in HeLa cultures). GSK-J4 treatment is shown to block LPS-induced TNF-α production in primary human macrophage cultures (IC50 = 9 µM by ELISA).


This probe is supplied in conjunction with the Structural Genomics Consortium (SGC). For further characterization details, please visit the SGC website (http://www.thesgc.org/scientists/chemical_probes/GSKJ1).
The H3K27me3 demethylase inhibitor GSK-J1 (Cat. No. 420204) against JMJD3 and UTX is made cell-permeable by ethyl ester derivatization to facilitate cellular delivery ([medium GSK-J4]t=0 = 30 µM; [cellular GSK-J1]t=1h = 1.6 µM). The pro-drug GSK-J4, being ineffective in cell-free assays, is effectively converted to GSK-J1 intracellularly and inhibits nuclear H3K27me3 levels in both untransfected and JMJD3-transfected HeLa cells (25 to 50 µM). Simultaneous inhibition of cellular JMJD3 and UTX by GSK-J4 treatment is shown to block LPS-induced production of several proinflammatory cytokines, including TNF-α (IC50 = 9 µM by ELISA), in primary human macrophage cultures.

Biochem/physiol Actions

Cell permeable: yes
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months for -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service