Skip to Content
Merck
All Photos(1)

Key Documents

5.00498

Sigma-Aldrich

HIF Inhibitor V

Synonym(s):

HIF Inhibitor V, Hypoxia-Inducible Factor Inhibitor V, ( E)-3-(3-(4-Adamantan-1-ylphenoxy)acrylamido)benzoic acid morpholinoethyl ester

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€213.00

€213.00


Estimated to ship on16 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€213.00

About This Item

Empirical Formula (Hill Notation):
C32H38N2O5
CAS Number:
Molecular Weight:
530.65
UNSPSC Code:
12352200
NACRES:
NA.77

€213.00


Estimated to ship on16 May 2025


Request a Bulk Order

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

storage temp.

2-8°C

Biochem/physiol Actions

Cell permeable: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.
Use only fresh DMSO for reconstitution.

Other Notes

Naik, R., et al. 2012. J. Med. Chem.55, 10564.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service