Skip to Content
Merck
All Photos(1)

Key Documents

499610

Sigma-Aldrich

(5Z)-7-Oxozeaenol, Curvularia sp.

(5Z)-7-Oxozeaenol, CAS 66018-38-0, is a cell-permeable, highly potent, selective, irreversible, and ATP-competitive inhibitor of MAPKKK TAK1 activity (IC₅₀ = 8 nM).

Synonym(s):

(5Z)-7-Oxozeaenol, Curvularia sp., TAK1 Inhibitor

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€101.00

€101.00


Estimated to ship on21 May 2025



Select a Size

Change View
1 MG
€101.00

About This Item

Empirical Formula (Hill Notation):
C19H22O7
CAS Number:
Molecular Weight:
362.37
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€101.00


Estimated to ship on21 May 2025


Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 10 mg/mL
ethanol: 5 mg/mL

shipped in

wet ice

storage temp.

−20°C

SMILES string

O1C(CC=CC(=O)C(C(CC=Cc2c(c(cc(c2)OC)O)C1=O)O)O)C

InChI

1S/C19H22O7/c1-11-5-3-7-14(20)18(23)15(21)8-4-6-12-9-13(25-2)10-16(22)17(12)19(24)26-11/h3-4,6-7,9-11,15,18,21-23H,5,8H2,1-2H3

InChI key

NEQZWEXWOFPKOT-UHFFFAOYSA-N

General description

A naturally isolated cell-permeable fungal resorcylic lactone that acts as a selective and highly potent inhibitor against the MAPKKK TAK1 activity (IC50 = 8 nM) in an ATP-competitive and irreversible manner, while inhibiting MEK1 (IC50 = 411 nM) and three other MAPKKKs (IC50 ≥268 nM against MEKK1, ASK1, and MEKK4) only at much higher concentrations. Shown to inhibit IL-1-induced/TAK1-mediated, but not H2O2-induced/TAK1-independent, JNK and p38 phosphorylation in 293 cells. (5Z)-Zeaenol, Curvularia sp. (Cat. No. 499609) may serve as a negative control.

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Windheim, M., et al. 2007. Biochem. J.404, 179.
Choo, M.K., et al. 2006. Mol. Cancer Ther.5, 2970.
Safwat, N., et al. 2005. Endocrinology146, 4814.
Ninomiya-Tsuji, J., et al. 2003. J. Biol. Chem.278, 18485.
Rawlins, P., et al. 1999. Int. J. Immunopharmacol.21, 799.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christoph Lammel et al.
Biomolecules, 10(9) (2020-08-23)
Twenty natural remedies traditionally used against different inflammatory diseases were probed for their potential to suppress the expression of the inflammatory markers E-selectin and VCAM-1 in a model system of IL-1 stimulated human umbilical vein endothelial cells (HUVEC). One third

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service