Skip to Content
Merck
All Photos(1)

Documents

48-690MAG

Millipore

MILLIPLEX MAP T-Cell Receptor Signaling Magnetic Bead Kit 7-Plex - Cell Signaling Multiplex Assay

Synonym(s):

7-Plex Magnetic Bead Kit, Magnetic Bead Kit, T-Cell Receptor Signaling Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.47

species reactivity

human

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

assay range

inter-assay cv: 4-17%
intra-assay cv: 3-8%

detection method

fluorometric (Luminex xMAP)

storage temp.

2-8°C

General description

The Milliplex® MAP 7-Plex Human T-Cell Receptor (TCR) Magnetic Bead Kit is used to detect changes in phosphorylated CREB (Ser133), Erk/MAP kinase 1/2 (Thr185/Tyr187), and phosphorylated tyrosine residues on CD3 epsilon chain, Lck, ZAP-70, LAT, and Syk in T-cell lysates using the Luminex® system. Although the protein kinase Syk is not part of the T-Cell Receptor pathway, it does play a significant role in B-Cell Receptor signaling and acts as a negative control for the analysis of T-Cell Receptor signaling. The detection assay is a rapid, convenient alternative to Western Blotting and immunoprecipitation procedures.

Specificity

Cross-reactivity between the antibodies and any of the other analytes in this panel is non-detectable or negligible.

Application

  • An overnight (4°C) incubation is recommended for best results.
  • This assay requires 25 μL diluted cell lysate.
  • This kit must be run using Assay Buffer 2 (provided).
  • 1 - 25 μg cell lysate/well (recommended starting concentration is 20 μg cell lysate/well).

Research Category
Cell Signaling
This cell signaling panel includes the following analytes in this multiplex kit: CD3ε (pan Tyr), CREB (S133), ERK (Thr185/Tyr187), LAT (pan Tyr), Lck (pan Tyr), Syk (pan Tyr), ZAP-70 (pan Tyr).
Used to detect/quantify:CD3ε (pan Tyr)|CREB (S133)|ERK (Thr185/Tyr187)|LAT (pan Tyr)|Lck (pan Tyr)|Syk (pan Tyr)|ZAP-70 (pan Tyr)

Storage and Stability

Recommended storage for kit components is 2 - 8°C

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

MILLIPLEX® cell signaling assays facilitate detection of phosphoproteins and total proteins in cellular pathways.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service