Skip to Content
Merck
All Photos(1)

Documents

48-602MAG

Millipore

MILLIPLEX® Cell Signaling Buffer and Detection Kit for Magnetic Beads

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.25

species reactivity

human

Quality Level

manufacturer/tradename

Milliplex®

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex® xMAP®)

storage temp.

2-8°C

General description

The MILLIPLEX® MAP Cell Signaling Buffer and Detection Kit is an optimized reagent system used for single target and multiplexing assays of MILLIPLEX® MAP Cell Signaling Magnetic Bead MAPmates. The Buffer and Detection Kit contains a set of common reagents that are used in conjunction with the Cell Signaling Magnetic Bead MAPmates for multiplex analysis of proteins in cell lysates using a Luminex 100 IS, 200, or HTS system. MAPmates are sold individually and consist of an antibody capture bead set, a biotinylated detection antibody and, as appropriate, a positive and negative control. The detection assay is a rapid, convenient alternative to Western Blotting and immunoprecipitation procedures. Each kit contains sufficient reagents for one 96 well assay plate.

Application

This MILLIPLEX® Cell Signaling Buffer & Detection Kit for Magnetic Beads is an optimized reagent system used for single target & multiplexing assays of MILLIPLEX MAP Cell Signaling Magnetic Bead MAPmates.

Packaging

96-well plate. Kit capacity: 100 assay points

Storage and Stability

Recommended storage for kit components is 2 - 8°C

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 2 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

MILLIPLEX® genotoxicity assays enable high-throughput measurement of DNA damage, reducing time and cost.

MILLIPLEX® cell signaling assays facilitate detection of phosphoproteins and total proteins in cellular pathways.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service