Skip to Content
Merck
All Photos(1)

Key Documents

481486

Sigma-Aldrich

NF-κB SN50M, Cell-Permeable Inactive Control Peptide

An inactive control for SN50 peptide.

Synonym(s):

NF-κB SN50M, Cell-Permeable Inactive Control Peptide, (AAVALLPAVLLALLAPVQRNGQKLMP)

Sign Into View Organizational & Contract Pricing

Select a Size

500 μG
€214.00

€214.00


Estimated to ship on22 May 2025


Request a Bulk Order

Select a Size

Change View
500 μG
€214.00

About This Item

Empirical Formula (Hill Notation):
C123H215N33O30S
Molecular Weight:
2668.29
UNSPSC Code:
12352200
NACRES:
NA.77

€214.00


Estimated to ship on22 May 2025


Request a Bulk Order

Quality Level

Assay

≥97% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

An inactive control for SN50 peptide (Cat. No. 481480). Corresponds to the SN50 peptide sequence with substitutions of Asn for Lys363 and Gly for Arg364 in the NLS region. In murine endothelial LE-II cells induced by LPS, this peptide had no measurable effect on NF-κB translocation at 18 µM.

Biochem/physiol Actions

Cell permeable: no
Primary Target
an inactive control for SN50 peptide
Product does not compete with ATP.
Reversible: no

Packaging

Yes

Warning

Toxicity: Standard Handling (A)

Sequence

H-Ala-Ala-Val-Ala-Leu-Leu-Pro-Ala-Val-Leu-Leu-Ala-Leu-Leu-Ala-Pro-Val-Gln-Arg-Asn-Gly-Gln-Lys-Leu-Met-Pro-OH

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C. Avoid/freeze thaw cycles of solutions.

Other Notes

Lin, Y.-Z., et al. 1995. J. Biol. Chem. 270, 14255.

Legal Information

Sold under license of U.S. Patents 5,807,746 and 6,043,339 for research use only. Licenses for commercial manufacture, commercial use, diagnostics, or therapeutics may be obtained by contacting Vanderbilt University, Nashville, Tennessee.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service