Skip to Content
Merck
All Photos(1)

Key Documents

480716

Sigma-Aldrich

α2-3,6,8,9-Neuraminidase, Arthrobacter ureafaciens, Recombinant, E. coli

Synonym(s):

α2-3,6,8,9-Neuraminidase, Arthrobacter ureafaciens, Recombinant, E. coli, N-Acetylneuraminyl Hydrolase, Sialidase

Sign Into View Organizational & Contract Pricing

Select a Size

250 MI.U.
€481.00

€481.00


Estimated to ship on25 May 2025



Select a Size

Change View
250 MI.U.
€481.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

€481.00


Estimated to ship on25 May 2025


recombinant

expressed in E. coli

Quality Level

description

RTECS - QQ3450000

form

liquid

specific activity

≥40 units/mg protein
≥5 units/mL

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

N-acetylglucosaminidase, α-fucosidase, α- and β-galactosidase, α- and β-mannosidase, proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Note: 1 mU = 1 milliunit.
Recombinant, Arthrobacter ureafaciens α2-3,6,8,9-Neuraminidase expressed in E. coli. Catalyzes the hydrolysis of all non-reducing terminal sialic acid residues from complex carbohydrates and glycoproteins. High concentrations of this sialidase and prolonged periods of incubation result in the additional cleavage of branched sialic acids. This enzyme is found as two active species of MW 60000 and 69000.

Warning

Toxicity: Harmful (C)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the release of 1 µmol of methylumbelliferone from 2ʹ-(4-methylumbelliferyl)-α-D-N-acetylneuraminic acid per min at 37°C, pH 5.0. (Note: 1 mU = 1 milliunit).

Physical form

In 20 mM Tris-HCl, 20 mM NaCl, pH 7.5.

Other Notes

Means, R.E. and Desrosiers, R.C. 2000. J. Virol.74, 11181.
Prime, S., et al. 1996. J. Chromatogr. A. 720, 263.
Roggentin, P., et al. 1995. Biol. Chem. Hoppe Seyler 376, 569
Dwek, R.A., et al. 1993. Annu. Rev. Biochem. 62, 65.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Karima S Khimani et al.
Scientific reports, 10(1), 2933-2933 (2020-02-23)
To compare goblet cell (GC) number and area in the covered superior (SB) versus exposed temporal (TB) bulbar conjunctiva in control versus aqueous tear deficient eyes (ATD) and evaluate correlation with tear MUC5AC protein. SB and TB impression cytology performed
Sarah W Nelson et al.
Influenza and other respiratory viruses, 13(6), 593-602 (2019-08-09)
The influenza A virus (IAV) binds to α-2,3- and α-2,6-linked sialic acid (SA) receptors expressed by Madin-Darby canine kidney (MDCK) cells. The receptor distribution may therefore be important in regulating IAV propagation. Serum-free medium (SFM) avoids variability in conventional culture

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service