Skip to Content
Merck

480708

Sigma-Aldrich

α2-3,6-Neuraminidase, Clostridium perfringens, Recombinant, E. coli

Synonym(s):

α2-3,6-Neuraminidase, Clostridium perfringens, Recombinant, E. coli, Acetylneuraminyl Hydrolase, Sialidase

Sign Into View Organizational & Contract Pricing

Select a Size

500 MI.U.
€445.00

€445.00


Estimated to ship on31 May 2025


Request a Sample

Select a Size

Change View
500 MI.U.
€445.00

About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.54

€445.00


Estimated to ship on31 May 2025


Request a Sample

recombinant

expressed in E. coli

Quality Level

description

RTECS - QQ3450000

product line

EMPROVE® EXPERT

form

liquid

specific activity

≥10 units/mL
≥250 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

foreign activity

N-acetylglucosaminidase, α-fucosidase, α- and β-galactosidase, α- and β-mannosidase, proteases, none detected

shipped in

wet ice

storage temp.

2-8°C

General description

Note: 1 mU = 1 milliunit.
Recombinant, Clostridium perfringens α2-3,6-Neuraminidase expressed in E. coli. Catalyzes the hydrolysis of non-reducing terminal α2,3 and α2,6 unbranched sialic acid residues from complex carbohydrates and glycoproteins. This enzyme does not exhibit activity on α2,8 or branched sialic acids.

Application

Recommended reaction buffer: 50 mM sodium phosphate buffer, pH 6.0

Warning

Toxicity: Harmful (C)

Unit Definition

One unit is defined as the amount of enzyme that will catalyze the release of 1 µmol of methylumbelliferone from 2ʹ-(4-Methylumbelliferyl)-α-D-N-acetylneuraminic Acid (Cat. No. 474495) per min at 37°C, pH 5.0.

Physical form

In 25 mM NaCl, 20 mM Tris-HCl, pH 7.5.

Other Notes

Prime, S., et al. 1996. J. Chromatogr. A. 720, 263.
Dwek, R.A., et al. 1993. Annu. Rev. Biochem. 62, 65.
Ohta, Y., et al. 1989. J. Biochem (Tokyo) 106, 1086.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
Emprove is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8B - Non-combustible, corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


What is the Emprove® Program?

The Emprove® Program is a system providing comprehensive and thorough documentation of our filters and single-use components, pharma raw materials, and starting materials. Four document types are included with an Emprove® Program subscription:

Material Qualification Dossier

Free of charge - Sign in to download

Quality Management Dossier (Open Access)

Free of charge - Sign in to download

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Operational Excellence Dossier

Available by subscription or for a fee

Learn more about the benefits an Emprove® Program subscription can provide
Why are these documents unavailable?
Emprove® Dossiers contain information intended to support the registration process and risk assessment activities of pharmaceutical producers and contain confidential information. Please contact us if you would like access.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yi Yang et al.
International journal of molecular sciences, 21(22) (2020-11-19)
Polysialic acid (polySia/PSA) is a linear homopolymer of sialic acid (Sia) that primarily modifies the neural cell adhesion molecule (NCAM) in mammalian brains. PolySia-NCAM not only displays an anti-adhesive function due to the hydration effect, but also possesses a molecule-retaining
Airi Mori et al.
International journal of molecular sciences, 21(16) (2020-08-23)
Polysialic acid (polySia/PSA) is an anionic glycan polymer of sialic acid, and it mostly modifies the neural cell adhesion molecule (NCAM) in mammalian brains. Quality and quantity of the polySia of the polySia-NCAM is spatio-temporally regulated in normal brain development

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service