Skip to Content
Merck
All Photos(1)

Key Documents

462711

Sigma-Aldrich

Methyl-α-D-mannopyranoside

Methyl-α-D-mannopyranoside, CAS 617-04-9, is a substrate for α1,2-Mannosyltransferase (Cat. No. 444125).

Synonym(s):

Methyl-α-D-mannopyranoside, α-Methyl-D-mannoside

Sign Into View Organizational & Contract Pricing

Select a Size

100 G
€144.00

€144.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
100 G
€144.00

About This Item

Empirical Formula (Hill Notation):
C7H14O6
CAS Number:
Molecular Weight:
194.18
MDL number:
UNSPSC Code:
12352201
NACRES:
NA.77

€144.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

Assay

≥99% (TLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

color

white to off-white

solubility

water: soluble

cation traces

heavy metals: ≤10 ppm

shipped in

ambient

storage temp.

10-30°C

SMILES string

O1C(C(C(C(C1CO)O)O)O)OC

InChI

1S/C7H14O6/c1-12-7-6(11)5(10)4(9)3(2-8)13-7/h3-11H,2H2,1H3

InChI key

HOVAGTYPODGVJG-UHFFFAOYSA-N

General description

Substrate for α1,2-Mannosyltransferase (Cat. No. 444125). Lectin inhibitory sugar.

Warning

Toxicity: Standard Handling (A)

Other Notes

Derewenda, Z., et al. 1989. EMBO J.. 8, 2189.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

404.1 °F

Flash Point(C)

206.74 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Z Derewenda et al.
The EMBO journal, 8(8), 2189-2193 (1989-08-01)
A complex of concanavalin A with methyl alpha-D-mannopyranoside has been crystallized in space group P212121 with a = 123.9 A, b = 129.1 A and c = 67.5 A. X-ray diffraction intensities to 2.9 A resolution have been collected on

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service