Skip to Content
Merck
All Photos(2)

Key Documents

444966

Sigma-Aldrich

MEK1/2 Inhibitor III

The MEK1/2 Inhibitor III, also referenced under CAS 391210-10-9, controls the biological activity of MEK1/2. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

MEK1/2 Inhibitor III, PD325901, PD0325901, N-((2R)-2,3-Dihyroxypropoxy)-3,4-difluoro-2-((2-fluoro-4-iodophenyl)amino)-benzamide, PD 0325901, MEK Inhibitor III

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€176.00

€176.00


Estimated to ship on29 May 2025



Select a Size

Change View
5 MG
€176.00

About This Item

Empirical Formula (Hill Notation):
C16H14F3IN2O4
CAS Number:
Molecular Weight:
482.19
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€176.00


Estimated to ship on29 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white to off-white

solubility

DMSO: 20 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Ic1cc(c(cc1)Nc2c(c(ccc2C(=O)NOC[C@H](O)CO)F)F)F

InChI

1S/C16H14F3IN2O4/c17-11-3-2-10(16(25)22-26-7-9(24)6-23)15(14(11)19)21-13-4-1-8(20)5-12(13)18/h1-5,9,21,23-24H,6-7H2,(H,22,25)/t9-/m1/s1

InChI key

SUDAHWBOROXANE-SECBINFHSA-N

General description

A cell-permeable benzamide compound that acts as a non-competitive, highly potent, selective MEK/MAPKK/MKK inhibitor and effectively blocks cellular Erk1/2 phosphorylation (by >90% in serum-starved HeLa after serum-stimulation and 50% in C26 with 25 and 0.33 nM inhibitor, respectively), while exhibiting little or no effect against the activity of Erk1/2 or a panel of 66 other kinases even at concentrations as high as 10 µM in cell-free kinase assays. Shown to be orally available in mice and suppress both the growth of and Erk phosphorylation in BRAF(V600E)-expressing SKMEL28 cell-derived tumor in vivo. Also available in InSolution format (Cat. No. 444968).

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Barrett, S.D., et al. 2008. Bioorg. Med. Chem. Lett.18, 6501.
Leyton, J., et al. 2008. Mol. Cancer Ther.7, 3112.
Silva, J., et al. 2008. PLoS Biol.6, 2237.
Ying, Q.L., et al. 2008. Nature453, 519.
Bain, J., et al. 2007. Biochem. J.408, 297.
Solit, D.B., et al. 2006. Nature439, 358.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phillip Wulfridge et al.
eLife, 10 (2021-02-24)
R-loops are three-stranded nucleic acid structures with essential roles in many nuclear processes. However, their unchecked accumulation is associated with genome instability and is observed in neurodevelopmental diseases and cancers. Genome-wide profiling of R-loops in normal and diseased cells can
Jian Yang et al.
Cell reports, 43(3), 113775-113775 (2024-02-21)
In mammals, many retrotransposons are de-repressed during zygotic genome activation (ZGA). However, their functions in early development remain elusive largely due to the challenge to simultaneously manipulate thousands of retrotransposon insertions in embryos. Here, we applied CRISPR interference (CRISPRi) to
Qingqing Yan et al.
Nature communications, 13(1), 53-53 (2022-01-12)
R-loops are three-stranded nucleic acid structures that accumulate on chromatin in neurological diseases and cancers and contribute to genome instability. Using a proximity-dependent labeling system, we identified distinct classes of proteins that regulate R-loops in vivo through different mechanisms. We
Ana Petracovici et al.
Molecular cell, 81(12), 2625-2639 (2021-04-23)
The Polycomb repressive complex 2 (PRC2) is an essential epigenetic regulator that deposits repressive H3K27me3. PRC2 subunits form two holocomplexes-PRC2.1 and PRC2.2-but the roles of these two PRC2 assemblies during differentiation are unclear. We employed auxin-inducible degradation to deplete PRC2.1
Oriol Iborra-Egea et al.
Methods in molecular biology (Clifton, N.J.), 2454, 61-81 (2021-12-01)
Stem cell therapy has an unparalleled potential to treat blood cancers, cardiovascular diseases and neurodegenerative conditions, among others. However, stem cell therapeutics must overcome multiple requirements before reaching clinical trials, including large animal safety and efficacy studies. In cardiovascular diseases

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service