Skip to Content
Merck
All Photos(1)

Key Documents

5.00496

Sigma-Aldrich

C59

≥99% (HPLC), powder, Porcn inhibitor, Calbiochem®

Synonym(s):

Porcn Inhibitor II, C59, NAN, Wnt-C59, Wnt Pathway Inhibitor XVI, 2-(4-(2-Methylpyridin-4-yl)phenyl)-N-(4-(pyridin-3-yl)phenyl)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€196.00

€196.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€196.00

About This Item

Empirical Formula (Hill Notation):
C25H21N3O
CAS Number:
Molecular Weight:
379.45
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€196.00


Estimated to ship on28 May 2025


Request a Bulk Order

Product Name

Porcn Inhibitor II, C59,

Assay

≥99% (HPLC)

Quality Level

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

off-white

solubility

DMSO: 100 mg/mL

storage temp.

2-8°C

SMILES string

N(c3ccc(cc3)c4cnccc4)C(=O)Cc1ccc(cc1)c2cc(ncc2)C

InChI

1S/C25H21N3O/c1-18-15-22(12-14-27-18)20-6-4-19(5-7-20)16-25(29)28-24-10-8-21(9-11-24)23-3-2-13-26-17-23/h2-15,17H,16H2,1H3,(H,28,29)

InChI key

KHZOJCQBHJUJFY-UHFFFAOYSA-N

General description

A cell-permeable pyridinylphenyl-acetamide that is much more potent than IWP-2 (Cat. No. 681671) in inhibiting MBOAT (membrane-bound O-acyltranferase) family member Porcn- (Porcupine) mediated Wnt palmitoylation (IC50 = 74 pM in STF reporter assays using Wnt3a-transfected HEK293 cells). Shown to be effective against Porcn of murine (mPorcn-A, B, C, D) and human (hPorcn-B), but not xenopus (xPorcn-A), species and potently inhibit the processing of both canonical (1, 2, 3a, 6, 7b, 8a, 9a, 9b, 10) and non-canonical (4, 5a, 11, 16) Wnt subtypes. C59 is demonstrated to be orally active and effectively suppress MMTV-WNT1 tumor expansion in mice (5 mg/kg & 10 mg/kg p.o.) in vivo.

Application

Porcn Inhibitor II, C59 has been used as a Wnt inhibitor in the differentiation protocol of hiPSCs to cardiomyocytes.[1]

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Porcupine (Porcn)-mediated Wnt palmitoylation
Reversible: yes

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alireza Rafieerad et al.
Advanced functional materials, 31(30), 2100015-2100015 (2022-03-11)
The application of nontoxic 2D transition-metal carbides (MXenes) has recently gained ground in bioelectronics. In group-4 transition metals, tantalum possesses enhanced biological and physical properties compared to other MXene counterparts. However, the application of tantalum carbide for bioelectrodes has not

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service