Skip to Content
Merck
All Photos(2)

Key Documents

438075

Sigma-Aldrich

Longdaysin

A cell-permeable purine compound that acts as a reversible and ATP-competitive dual inhibitor of CKIα and CKIδ activities with moderate selectivity over Cdk7 and Erk2.

Synonym(s):

Longdaysin, 9-Isopropyl-N-(3-(trifluoromethyl)benzyl)-9H-purin-6-amine, Casein Kinase I Inhibitor VII, Casein Kinase I Inhibitor VII, 9-Isopropyl-N-(3-(trifluoromethyl)benzyl)-9H-purin-6-amine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16F3N5
CAS Number:
Molecular Weight:
335.33
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥99% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 50 mg/mL

shipped in

wet ice

storage temp.

2-8°C

General description

A cell-permeable purine compound that acts as a reversible and ATP-competitive dual inhibitor of CKIα and CKIδ activities (IC50 = 5.6 and 8.8 µM) with moderate selectivity over Cdk7 and Erk2 (IC50 = 29 and 52 µM). Shown to block CKIα/δ-mediated phosphorylation of PER1, a clock protein, (IC50 ~ 9 µM) and repress PER1 proteasomal degradation, and lengthen the circadian period in U2OS cells (by ~ 13 hrs at 10 µM) much more effectively than Casein Kinase I Inhibitor, D4476 (Cat. No. 218696) and IC261 (Cat. No. 400090) and in larval zebra fish.

Packaging

Packaged under inert gas

Warning

Toxicity: Harmful (C)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Hirota, T., et al. 2010. PLoS Biol.8, e1000559.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service