Skip to Content
Merck
All Photos(1)

Key Documents

4310-OP

Millipore

Ethidium Bromide

OmniPur® Grade, >=98.0%, Fluorescent stain

Synonym(s):

OmniPur® Ethidium Bromide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H20BrN3
CAS Number:
Molecular Weight:
394.31
MDL number:
UNSPSC Code:
41105322

Quality Level

form

crystalline powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

dark purple

shipped in

ambient

InChI

1S/C21H19N3.BrH/c1-2-24-20-13-16(23)9-11-18(20)17-10-8-15(22)12-19(17)21(24)14-6-4-3-5-7-14;/h3-13,23H,2,22H2,1H3;1H

InChI key

ZMMJGEGLRURXTF-UHFFFAOYSA-N

General description

Fluorescent stain for illustrating nucleic acids in gels. Facilitates purification of DNA in Cesium Chloride gradient. Suitable for fluorescent staining of nucleic Acids. Molecular biology grade.

Specifications:
Assay (dry basis): ≥98.0%
DNase: None detected
Loss on drying: ≤8.0%
Protease: None detected
RNase: None detected





Intended for laboratory and manufacturing use only. Not for drug, food, or household use.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany
OMNIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Oral - Muta. 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service