Skip to Content
Merck
All Photos(1)

Key Documents

382180

Sigma-Aldrich

Histone Deacetylase 6, GST-Fusion, Human Recombinant, S. frugiperda

Histone Deacetylase 6, GST-Fusion, Human Recombinant, S. frugiperda, is a full-length HDAC6 fused to GST at the N-terminus. Useful in the study of HDAC6 regulation and for inhibitor screening.

Synonym(s):

HDAC6, Human, Recombinant

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
€538.00

€538.00


Estimated to ship on23 May 2025



Select a Size

Change View
50 μG
€538.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.54

€538.00


Estimated to ship on23 May 2025


recombinant

expressed in insect cells (S. frugiperda)

Quality Level

Assay

≥80% (SDS-PAGE)

form

liquid

specific activity

≥50 units/μg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Full length, recombinant, human histone deacetylase 6 (HDAC6) fused to GST at the N-terminus and expressed in S. frugiperda insect cells using a baculovirus expression system. Actively maintained in the cytoplasm, HDAC6 is uniquely dependent on its ubiquitin-binding domain and functions at the nexus between two cellular signaling systems, involving protein acetylation and ubiquitination. The active deacetylase is useful in the study of HDAC6 regulation and for inhibitor screening.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enyme that is required to deacetylate 1 pmol fluorescent HDAC peptide substrate per min at 30°C.

Physical form

40 mM Tris-HCl pH8.0, 110mM NaCl, 2.2 mM KCl, 0.04% Tween 20, 3mM DTT, and 20% glycerol.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Pandey, U.B., et al. 2007. Nature447, 859.
Hubbert, C., et al. 2002. Nature417, 455.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin Kyun Park et al.
Arthritis research & therapy, 23(1), 177-177 (2021-07-07)
To investigate the effects of inhibiting histone deacetylase (HDAC) 6 on inflammatory responses and tissue-destructive functions of fibroblast-like synoviocytes (FLS) in rheumatoid arthritis (RA). FLS from RA patients were activated with interleukin (IL)-1β in the presence of increasing concentrations of
Nina Ha et al.
British journal of pharmacology, 177(22), 5096-5113 (2021-01-19)
Charcot-Marie-Tooth (CMT) disease is the most common hereditary peripheral neuropathy. CMT type 1A (CMT1A) accounts for approximately 50% of CMT patients and is linked to PMP22 gene duplication. Histone deacetylase-6 (HDAC6) has pleiotropic effects, such as regulating lipid homeostasis and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service