Merck
  • A novel histone deacetylase 6 inhibitor improves myelination of Schwann cells in a model of Charcot-Marie-Tooth disease type 1A.
A novel histone deacetylase 6 inhibitor improves myelination of Schwann cells in a model of Charcot-Marie-Tooth disease type 1A.
British journal of pharmacology (2021-01-19)
Nina Ha, Young Il Choi, Namhee Jung, Ju Young Song, Dae Kwon Bae, Min Cheol Kim, Yong Jae Lee, Hyeseung Song, Geon Kwak, Soyeon Jeong, Saeyoung Park, Soo Hyun Nam, Sung-Chul Jung, Byung-Ok Choi
ABSTRACT
Charcot-Marie-Tooth (CMT) disease is the most common hereditary peripheral neuropathy. CMT type 1A (CMT1A) accounts for approximately 50% of CMT patients and is linked to PMP22 gene duplication. Histone deacetylase-6 (HDAC6) has pleiotropic effects, such as regulating lipid homeostasis and cellular stress. Although HDAC6 has been regarded as a promising drug target for neurodegenerative diseases, its inhibition has not yet been tested in CMT1A. Here we have tested the therapeutic potential of CKD-504, a clinical stage HDAC6 inhibitor, in a mouse model of CMT1A EXPERIMENTAL APPROACH: The potency and selectivity of CKD-504 was evaluated, using a HDAC enzyme panel assay and western blots. The therapeutic potential of CKD-504 was evaluated using behavioural testing and electrophysiological assessments in the C22 mouse model of CMT1A. PMP22 protein expression and aggregation were analysed in mesenchymal stem cell-derived Schwann cells from CMT1A patients and sciatic nerves from C22 mice. The HDAC6 inhibitor, CKD-504, modulated molecular chaperon proteins such as HSP90 and HSP70, which are involved in the folding/refolding of proteins such as PMP22. CKD-504 treatment restored myelination in both mesenchymal stem cell-derived Schwann cells from CMT1A patients and sciatic nerves of C22 mice and improved the axonal integrity of the sciatic nerve, leading to behavioural, electrophysiological, and histological improvements in C22 mice. A novel HDAC6 inhibitor, CKD-504, has potent therapeutic efficacy for CMT1A.
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-LAMP1 antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-GAPDH antibody produced in rabbit, ~1 mg/mL, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Anti-Mitochondria Antibody, surface of intact mitochondria, clone 113-1, clone 113-1, Chemicon®, from mouse
Sigma-Aldrich
Anti-Myelin Basic Protein Antibody, Chemicon®, from chicken
Sigma-Aldrich
Histone Deacetylase 6, GST-Fusion, Human Recombinant, S. frugiperda, Histone Deacetylase 6, GST-Fusion, Human Recombinant, S. frugiperda, is a full-length HDAC6 fused to GST at the N-terminus. Useful in the study of HDAC6 regulation and for inhibitor screening.
Roche
cOmplete, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks