Skip to Content
Merck
All Photos(1)

Key Documents

373272

Millipore

HECAMEG

Non-ionic detergent useful for solubilization of membrane-bound proteins in their native state. Easily removed by dialysis.

Synonym(s):

Methyl-6-O-(N-heptylcarbamoyl)-α-D-glucopyranoside

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€398.00

€398.00


Estimated to ship on25 May 2025


Request a Bulk Order

Select a Size

Change View
5 G
€398.00

About This Item

Empirical Formula (Hill Notation):
C15H29NO7
CAS Number:
Molecular Weight:
335.39
MDL number:
UNSPSC Code:
12161902
NACRES:
NA.21

€398.00


Estimated to ship on25 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

crystalline solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

white

CMC

19.5 mM

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

15-25°C

SMILES string

N(CCCCCCC)C(=O)OC[C@H]1O[C@@H]([C@@H]([C@H]([C@@H]1O)O)O)OC

InChI

1S/C15H29NO7/c1-3-4-5-6-7-8-16-15(20)22-9-10-11(17)12(18)13(19)14(21-2)23-10/h10-14,17-19H,3-9H2,1-2H3,(H,16,20)/t10-,11-,12+,13-,14+/m1/s1

InChI key

XPIVOYOQXKNYHA-RGDJUOJXSA-N

General description

Non-ionic detergent useful for solubilization of membrane-bound proteins in their native state. Easily removed by dialysis.
Nonionic detergent useful for solubilization of membrane-bound proteins in their native state. Easily removed by dialysis. Has a CMC of 19.5 mM.

Application


  • An assessment of the biochemical applications of the non-ionic surfactant Hecameg.: This foundational study explores the biochemical applications of Hecameg, particularly its efficacy in solubilizing membrane proteins without denaturation, making it a valuable tool in membrane biochemistry research (Ruiz et al., 1994).

  • Purification and characterization of the cytochrome b6 f complex from Chlamydomonas reinhardtii.: Hecameg is utilized in this study for the purification of the cytochrome b6 f complex, demonstrating its effectiveness in maintaining the functional integrity of membrane protein complexes during isolation (Pierre et al., 1995).

  • Development of an improved in vitro activity assay for medium chain length PHA polymerases based on CoenzymeA release measurements.: This article describes the use of Hecameg in an improved assay for polymerase activity, emphasizing its role in biochemical methods that require the stabilization of enzyme activities (de Roo et al., 2000).

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, store in the refrigerator (4°C). Stock solutions are stable for up to 3 months at 4°C.

Other Notes

Plusquellec, D., et al. 1989. Anal. Biochem.179, 145.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service