Skip to Content
Merck
All Photos(1)

Key Documents

371207

Sigma-Aldrich

Dkk-1, Mouse, Recombinant

Dkk-1, Mouse, Recombinant, consists of amino acids 30-272 and expressed in Chinese Hamster Ovary cells. Inhibits Wnt-3a-induced alkaline phosphatase production by MC3T3-E1 mouse osteoblastic cells.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€565.00

€565.00


Estimated to ship on15 May 2025



Select a Size

Change View
10 μG
€565.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€565.00


Estimated to ship on15 May 2025


recombinant

expressed in CHO cells

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

impurities

<1.0 EU/μg protein Endotoxin

shipped in

ambient

storage temp.

−20°C

General description

Recombinant, mouse Dickkopf related protein 1 (Dkk-1) consisting of amino acids 30-272 and expressed in Chinese Hamster Ovary cells. Inhibits Wnt-3a-induced alkaline phosphatase production by MC3T3-E1 mouse osteoblastic cells. ND50 = 0.02-0.1 µg/ml.

Biochem/physiol Actions

ND₅₀ = 0.02-0.1 µg/ml, in the presence of 10 ng/ml of rmWnt-3a, as measured by its ability to inhibit Wnt-3a-inducedalkaline phosphatase production by MC3T3-E1 mouse osteoblastic cells.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from 0.2 µm sterile-filtered PBS, BSA.

Reconstitution

Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (-20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at -20°C.
Reconstitute to 100 µg/ml in PBS containing at least 0.1% human or bovine serum albumin.

Other Notes

Pinzone, J.J., et al. 2009. Blood113, 517.
Li, J., et al. 2006. Bone36, 754.
Morvan, F., et al. 2006. J. Bone Miner. Res.21, 934.
Niehrs, C., 2006. Oncogene25, 7469.
Verani, R., et al. 2006. J. Neurochem.101, 242.
Cappuccio, I., et al. 2005. J. Neurosci.25, 2647.
Kemp, C., et al. 2005. Dev. Dyn.233, 1064.
Adamska, M., et al. 2004. Dev. Biol.272, 134.
Bullock, C.M., et al. 2004. Mol. Pharmacol.65, 582.
Mao, B., et al. 2002. Nature417, 664.
Mao, B., et al. 2001. Nature411, 321.
Glinka, A., et al. 1998. Nature391, 357.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service