Skip to Content
Merck
All Photos(1)

Key Documents

370677

Sigma-Aldrich

Guanosine 3′,5′-cyclic Monophosphorothioate, 8-(4-Chlorophenylthio)-, Rp-Isomer, Triethylammonium Salt

A potent, cell-permeable, and metabolically-stable inhibitor of protein kinase G Iα, Iβ, and type II.

Synonym(s):

Guanosine 3′,5′-cyclic Monophosphorothioate, 8-(4-Chlorophenylthio)-, Rp-Isomer, Triethylammonium Salt, Rp-8- pCPT-cGMPS, TEA, PKG Inhibitor III

Sign Into View Organizational & Contract Pricing

Select a Size

1 μMOL
€325.00

€325.00


Estimated to ship on17 May 2025



Select a Size

Change View
1 μMOL
€325.00

About This Item

Empirical Formula (Hill Notation):
C16H15ClN5O6PS2 · xC6H15N
Molecular Weight:
503.88 (free acid basis)
UNSPSC Code:
41106305
NACRES:
NA.77

€325.00


Estimated to ship on17 May 2025


Quality Level

Assay

≥99% (HPLC)

form

lyophilized

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

solubility

water: 0.5 mg/mL
DMSO: soluble

shipped in

ambient

storage temp.

−20°C

General description

A potent, cell-permeable inhibitor of protein kinase G Iα, Iβ, and type II. A combination of the protein kinase inhibitor Rp-cGMPS and the widely used cGMP analog, 8-pCPT-cGMP. Significantly more lipophilic and membrane-permeant than Rp-cGMPS and Rp-8-Br-cGMPS. Resistant to hydrolysis by mammalian cyclic nucleotide dependent phosphodiesterases. Note: 1 µmol = 0.61 mg.
A potent, cell-permeable, and metabolically-stable inhibitor of protein kinase G Iα, Iβ, and type II. Significantly more lipophilic and membrane-permeable than Rp-cGMPS or Rp-8-Br-cGMPS (Cat. No. 370674). Note: 1 µmol = 0.61 mg.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
PKG1α
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Abd Alla, S., et al. 1996. Eur. J. Biochem.241, 498.
Lee, S.J., et al. 1996. Dev. Biol.180, 324.
Van Uffelen, B.E., et al. 1996. J. Leukoc. Biol.60, 94.
Willmott, N., et al. 1996. J. Biol. Chem.271, 3699.
Butt, E., et al. 1994. Eur. J. Pharmacol. 269, 265.

Legal Information

Sold under license of U.S. Patent 5,625,056, Patents DE 3,802,865.4, and DE 4,217,679 issued to BIOLOG LSI.
CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service