Skip to Content
Merck
All Photos(1)

Key Documents

116812

Sigma-Aldrich

Adenosine 3′,5′-cyclic Monophosphate, 8-(4-Chlorophenylthio)-, Sodium Salt

Cell-permeable cAMP analog that activates both protein kinase A and protein kinase G.

Synonym(s):

Adenosine 3′,5′-cyclic Monophosphate, 8-(4-Chlorophenylthio)-, Sodium Salt, Adenosine 3ʹ,5ʹ-cyclic Monophosphate, 8-(4-Chlorophenylthio)-, Sodium Salt

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
€58.60

€58.60


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 MG
€58.60

About This Item

Empirical Formula (Hill Notation):
C16H14ClN5O6PS · Na
CAS Number:
Molecular Weight:
493.79
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.77

€58.60


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

Assay

≥98% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: 10 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

[Na+].[P]1(=O)(O[C@@H]2[C@H](O[C@H]([C@@H]2O)[n]3c4ncnc(c4nc3Sc5ccc(cc5)Cl)N)CO1)[O-]

InChI

1S/C16H15ClN5O6PS.Na/c17-7-1-3-8(4-2-7)30-16-21-10-13(18)19-6-20-14(10)22(16)15-11(23)12-9(27-15)5-26-29(24,25)28-12;/h1-4,6,9,11-12,15,23H,5H2,(H,24,25)(H2,18,19,20);/q;+1/p-1/t9-,11-,12-,15-;/m1./s1

InChI key

YIJFVHMIFGLKQL-DNBRLMRSSA-M

Related Categories

General description

Cell-permeable cAMP analog that activates both protein kinase A and protein kinase G. Induces the guanine-nucleotide-exchange factor activity of EGFI, also known as Epac (exchange protein directly activated by cAMP), and EGFII towards Rap1 both in vivo and in vitro. Also acts as a potent inhibitor of cGMP specific phosphodiesterase (IC50 = 900 nM). Also inhibits cAMP-specific phosphodiesterases at much higher concentrations (IC50 = 25 µM).

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Protein kinase A and protein kinase G
Product does not compete with ATP.
Reversible: no
Target IC50: 900 nM against cGMP-specific phosphodiesterase

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 1 month at -20°C

Other Notes

de Rooij, J., et al. 1998. Nature.396, 474.
Kawasaki, H., et al. 1998. Science.282, 2275.
Moed, P.J., et al. 1993. Comp. Biochem. Physiol.105A, 711.
Connolly, B.J., et al. 1992. Biochem. Pharmacol.44, 2303.
Phillips, M.E., and Taylor, A. 1992. J. Physiol.456, 591.
Shipston, M.J., and Antoni, F.A. 1992. Endocrinology130, 2213.
Datta, R., et al. 1991. Blood78, 83.
Peters, D.J.M., et al. 1991. Proc. Natl. Acad. Sci. USA88, 9219.
Sandberg, M., et al. 1991. Biochem. J.279, 521.
Unterman, T.G., et al. 1991. Endocrinology128, 2693.
Xu, H., et al. 1989. Brain Res.504, 36.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paula V Monje
Bio-protocol, 13(19), e4840-e4840 (2023-11-30)
This paper introduces simple analytical methods and bioassays to promptly assess the identity and function of in vitro cultured human Schwann cells (hSCs). A systematic approach is proposed to unequivocally discriminate hSCs from other glial cells, non-glial cells, and non-human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service