Skip to Content
Merck
All Photos(1)

Key Documents

345831

Sigma-Aldrich

Galectin-1, Human, Recombinant, E. coli

Synonym(s):

Lactose-binding lectin 1, Beta-galactoside-binding lectin L-14-I, S-Lac lectin 1, Galaptin, 14 kDa lectin, HPL, HBL, MAPK activating protein MP12

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202

Assay

≥98% (SDS-PAGE)

Quality Level

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

solubility

water: 0.1-1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

Recombinant human galectin-1 expressed in and purified from E. coli. Galectins are a family of evolutionarily conserved animal lectins with affinity for β-galactosides that have homologous carbohydrate recognition domains (CRDs). Galectin-1 can induce apoptosis of activated T cells and leukemic T cell lines. It also binds to CD45 and inhibits its protein phosphatase activity.
Recombinant, human galectin-1 expressed in E. coli. Galectins are a family of evolutionarily conserved animal lectins with affinity for β-galactosides that have homologous carbohydrate recognition domains (CRDs). Galectin-1 can induce apoptosis of activated T cells and leukemic T cell lines. It also binds to CD45 and inhibits its protein phosphatase activity.

Warning

Toxicity: Standard Handling (A)

Physical form

Lyophilized from sterile-filtered 20 mM Tris-HCl, pH 7.5.

Preparation Note

Further dilute into aqueous buffers as needed.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C. Diluted working solutions are stable for up to 1 week at 4°C or for up to 1 month at -20°C.

Other Notes

Rabinovich, A., et al. 2002. Trends Immunol.23, 313.
Rabinovich, A., et al. 2002. J. Leukoc. Biol.71, 741.
Hughes, R.C., et al. 2001. Biochimie.83, 667.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service