Skip to Content
Merck
All Photos(1)

Key Documents

344291

Sigma-Aldrich

Freund′s Incomplete Adjuvant, Modified

Freund’s Incomplete Adjuvant, Modified Contains 85% Drakeol 5NF, 15% Arlacel A. Heat sterilized. Contains zero colony forming units.

Synonym(s):

Incomplete Freund′s Adjuvant

Sign Into View Organizational & Contract Pricing

Select a Size

10 ML
€73.10

€73.10


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
10 ML
€73.10

About This Item

UNSPSC Code:
41116133
NACRES:
NA.41

€73.10


Estimated to ship on29 May 2025


Request a Bulk Order

Quality Level

form

liquid (oily)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

yellow

shipped in

ambient

storage temp.

10-30°C

General description

Contains 85% Drakeol 5NF, 15% Arlacel A (mannide monooleate emulsifier). Heat sterilized. Contains zero colony forming units/ml.
The word "adjuvant" is derived from the Latin, adjuvare, meaning "to help." During immunization, an adjuvant is generally mixed with the antigen to enhance the rate and amount of antibody formed. The most popular complete adjuvant is that of Freund. It consists of killed and dried microbial cells suspended in an emulsifying oil. Adjuvant without the microbial cells is called "incomplete adjuvant." Complete adjuvant forms a stable water-in-oil emulsion with aqueous solution of antigen. When injected subcutaneously into animals, the emulsified antigen forms a depot from which antigen is delivered slowly and at sustained levels to the animal′s antibody-synthesizing system. The induced antibody response is usually high and sustained. In guinea pigs, an additional benefit from adjuvant is the delayed appearance of hypersensitivity.

Components

10 ml of Freund′s Incomplete Adjuvant homogenized in the following amounts by weight:

85% Drakeol 5 NF (light mineral oil)

15% Arlacel A, mannide monooleate emulsifier

Warning

Toxicity: Standard Handling (A)

Preparation Note

1. Take the vial of Friend′s Incomplete Adjuvant and fold the aluminum seal back to permit free access to the stopper. Be careful not to break the aluminum ring that holds the stopper in place.

2. Dissolve 50 mg of antigen in 5 ml sterile PBS, pH 7.0-7.3. Maintain or create sterility if at all feasible.

3. Draw the solution into a 10 ml syringe with a 1 inch 21 gauge needle.

4. Insert needle into adjuvant vial and forcibly drive solution into the vial. Withdraw about half the mixture into syringe and force it into the adjuvant vial again.

5. Repeat the mixing procedure in step 4 until a smooth, stable, milky, water-in-oil emulsion forms. Check if the emulsion is complete by expelling a drop from the syringe onto the water surface in a beaker. If the drop remains on the surface and does not disperse when the beaker is agitated slightly, the emulsion is adequate. A properly prepared emulsion is stable for days.

Other Notes

10 ml of Freund′s Incomplete Adjuvant homogenized in the following amounts by weight:



85% Drakeol 5 NF (light mineral oil)

15% Arlacel A, mannide monooleate emulsifier.



1. Take the vial of Friend′s Incomplete Adjuvant and fold the aluminum seal back to permit free access to the stopper. Be careful not to break the aluminum ring that holds the stopper in place.

2. Dissolve 50 mg of antigen in 5.0 ml of sterile PBS, pH 7.0-7.3. Maintain or create sterility if at all feasible.

3. Draw the solution into a 10 ml syringe with a 1 inch 21 gauge needle.

4. Insert needle into adjuvant vial and forcibly drive solution into the vial. Withdraw about half the mixture into syringe and force it into the adjuvant vial again.

5. Repeat the mixing procedure in step 4 until a smooth, stable, milky, water-in-oil emulsion forms. Check if the emulsion is complete by expelling a drop from the syringe onto the water surface in a beaker. If the drop remains on the surface and does not disperse when the beaker is agitated slightly, the emulsion is adequate. A properly prepared emulsion is stable for days.
Freund, J. 1947. Ann. Rev. Microbiol.1, 291.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Hello, I see that product #344291-10ML is heat sterilized. What temperature is it sterilized at? If possible, do you know if it would be able to withstand a second exposure at that temperature?

    1 answer
    1. Upon verification, our team indicated that this product was autoclaved three times at 121°C for 15 minutes each cycle.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service