Skip to Content
Merck
All Photos(1)

Key Documents

344289

Sigma-Aldrich

Freund′s Complete Adjuvant, Modified, Mycobacterium butyricum

This Freund’s Complete Adjuvant, Modified, Mycobacterium butyricum is validated for use in Immunization.

Synonym(s):

Freund’s Complete Adjuvant, Modified, Mycobacterium butyricum

Sign Into View Organizational & Contract Pricing

Select a Size

1 SET
€259.00

€259.00


Estimated to ship on28 May 2025


Request a Bulk Order

Select a Size

Change View
1 SET
€259.00

About This Item

CAS Number:
MDL number:
UNSPSC Code:
41116133
NACRES:
NA.41

€259.00


Estimated to ship on28 May 2025


Request a Bulk Order

description

RTECS - LS6860000

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

color

clear to slightly cloudy amber

shipped in

ambient

storage temp.

2-8°C

General description

1 Set (10 x 10 ml) each containing yellow, oily, bacterial suspension.
Contains 10 vials per set, each containing 10 ml of Freund′s complete adjuvant homogenized in the following proportions by weight: 85% Drakeol 5NF, 15% Arlacel A (mannide monooleate emulsifier), and 0.1% Mycobacterium butyricum dry cells.

Application

Immunization

Warning

Toxicity: Standard Handling (A)

Physical form

Yellow, oily, bacterial suspension.

Other Notes

Composition

The Freund′s Adjuvant set contains 10 identical ampules, each with 10 ml of Freund′s Complete Adjuvant homogenized in the following preparations by weight:

85% Drakeol 5 NF

15% Arlacel A, mannide monooleate emulsifier

0.1% Mycobacterium butyricum dry cells



Sample Preparation

1. Withdraw contents of ampule using a 10 ml syringe with a 1 inch 21 guage needle. Add to a 50 ml vial, preferably with a narrow neck.

2. Dissolve 50 mg of antigen in 5 ml sterile PBS, pH 7.0-7.3. Maintain or create sterility if at all feasible. Draw the solution into the syringe.

3. Insert needle into adjuvant vial and forcibly drive solution into the vial. Withdraw about half the mixture into syringe and force it into the adjuvant vial again.

4.Repeat the mixing procedure in step 4 until a smooth, stable, milky, water-in-oil emulsion forms. Mixture may seem to thicken as the emulsion forms. Check if the emulsion is complete by expelling a drop from the syringe onto the water surface in a beaker. If the drop remains on the surface and does not disperse when the beaker is agitated slightly, the emulsion is adequate. A properly prepared emulsion is stable for days.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Asp. Tox. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service