Skip to Content
Merck
All Photos(1)

Key Documents

344152

Sigma-Aldrich

FPT Inhibitor II

The FPT Inhibitor II controls the biological activity of FPT. This small molecule/inhibitor is primarily used for Cancer applications.

Synonym(s):

FPT Inhibitor II, (E,E)-2-[2-Oxo-2-[[(3,7,11-trimethyl-2,6,10-dodecatrienyl)oxy]amino]ethyl]phosphonic Acid, 2Na

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€162.00

€162.00


Estimated to ship on18 May 2025



Select a Size

Change View
1 MG
€162.00

About This Item

Empirical Formula (Hill Notation):
C17H28NO5P · 2Na
Molecular Weight:
403.36
UNSPSC Code:
12352200
NACRES:
NA.77

€162.00


Estimated to ship on18 May 2025


Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)

color

white

solubility

water: ≥10 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A cell permeable potent and selective inhibitor of farnesyltransferase (FTase; IC50 = 75 nM). Also inhibits Ras farnesylation in whole cells by ~90% at 25-250 µM. Resistant to cleavage by phosphorylation.

Please note that the molecular weight for this compound is batch-specific due to variable water content. Please refer to the vial label or the certificate of analysis for the batch-specific molecular weight. The molecular weight provided represents the baseline molecular weight without water.
A highly selective and potent inhibitor of Ras farnesyltransferase (IC50 = 75 nM). Inhibits geranylgeranyltransferase I (IC50 = 24 µM) and II (IC50 = 24 µM) at much higher concentrations. Also inhibits Ras farnesylation in whole cells by ~90% at 10-100 µg/ml (25-250 µM). Resistant to cleavage by phosphorylation.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no
Target IC50: 75 nM against Ftase

Warning

Toxicity: Standard Handling (A)

Other Notes

Manne, V., et al. 1995. Drug Development Res.34, 121.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David J Carroll et al.
Methods in molecular biology (Clifton, N.J.), 518, 57-66 (2008-12-17)
The starfish oocyte has proven useful for studies involving microinjection because it is relatively large (190 mum) and optically clear. These oocytes are easily obtained from the ovary arrested at prophase of meiosis I, making them useful as a model

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service