Skip to Content
Merck
All Photos(1)

Key Documents

234405

Sigma-Aldrich

Complement C5-Depleted Serum, Human

Synonym(s):

Complement Serum

Sign Into View Organizational & Contract Pricing

Select a Size

1 ML
€336.00

€336.00


Estimated to ship on26 May 2025


Request a Bulk Order

Select a Size

Change View
1 ML
€336.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.25
form:
liquid
shipped in:
dry ice

€336.00


Estimated to ship on26 May 2025


Request a Bulk Order

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

impurities

C5 antigen, none detected (Ouchterlony)

shipped in

dry ice

storage temp.

−70°C

General description

Human serum depleted of complement component C5 by affinity chromatography.
Human serum that has been depleted of complement component C5 by affinity chromatography. Intended for the detection and quantitation of hemolytically active C5 complement component. The classical and alternative pathways are intact except for the specifically depleted component. Recommended assay volume: ≤60 µl

Biochem/physiol Actions

Complement C5-depleted serum from humans produced by affinity chromatography aids the development of the complement cascade up to the desired convertase level. It eliminates the usual time-consuming, expensive, and stepwise formation of enzymatic complexes from purified products. Whole serum usage reduces the risk of protein inactivation during the process. It also promotes better physiological conditions.[1] Complement C5-Depleted Serum is intended for the detection and quantitation of the hemolytically active C5 complement component. Creating complement convertases on erythrocytes enhances a better readout of the assay by spectrophotometric quantification of released hemoglobin.[1]

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 145 mM NaCl, 10 mM sodium phosphate, pH 7.3.

Preparation Note

Prepared from plasma of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Marcin Okroj et al.
PloS one, 7(10), e47245-e47245 (2012-10-17)
C3 and C5 convertases are central stages of the complement cascade since they converge the different initiation pathways, augment complement activation by an amplification loop and lead to a common terminal pathway resulting in the formation of the membrane attack

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service