Skip to Content
Merck
All Photos(2)

Key Documents

219556

Sigma-Aldrich

Ceranib-2

A cell-permeable, potent inhibitor of ceramidase (IC₅₀ = 28 mM) with anti-proliferative and proapoptotic properties.

Synonym(s):

Ceranib-2, Ceranib 2, Ceranib2, 3-(3-(4-methoxyphenyl)acryloyl)-4-phenylquinolin-2(1H)-one, AC Inhibitor III, Acid Ceramidase Inhibitor III

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H19NO3
Molecular Weight:
381.42
MDL number:
UNSPSC Code:
12352200

Quality Level

Assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

white

solubility

DMSO: 100 mg/mL, clear, pale yellow

shipped in

ambient

storage temp.

−20°C

SMILES string

O=C(N1)C(C(/C=C/C2=CC=C(OC)C=C2)=O)=C(C3=CC=CC=C3)C4=C1C=CC=C4

General description

A cell-permeable, potent inhibitor of ceramidase (IC50 = 28 µM) with anti-proliferative and proapoptotic properties. Shown to block the proliferation of SKVO3, ovarian adenocarcinma cells (IC50 = 730 nM) in the G2/M phase. When used in combination with paclitaxel, it increases tumor chemosentitvity and exhibits additive chemotherapeutic effects. Reduces tumor burden in Balb/c mice when administered intraperitoneally (20-50 mg/kg) without affecting the total body weight.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Draper, J., et al. 2011, Mol. Cancer Ther.10, 2052.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service