Skip to Content
Merck
All Photos(4)

Key Documents

WH0000581M1

Sigma-Aldrich

Monoclonal Anti-BAX antibody produced in mouse

clone 1F5-1B7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BCL2-associated X protein, Anti-Bax zeta

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
€418.00

€418.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1103


Select a Size

Change View
100 μG
€418.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

€418.00


Check Cart for Availability
A recombinant, preservative-free antibody is available for your target. Try ZRB1103

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1F5-1B7, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
indirect ELISA: suitable
proximity ligation assay: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BAX(581)

General description

The protein encoded by this gene belongs to the BCL2 protein family. BCL2 family members form hetero- or homodimers and act as anti- or pro-apoptotic regulators that are involved in a wide variety of cellular activities. This protein forms a heterodimer with BCL2, and functions as an apoptotic activator. This protein is reported to interact with, and increase the opening of, the mitochondrial voltage-dependent anion channel (VDAC), which leads to the loss in membrane potential and the release of cytochrome c. The expression of this gene is regulated by the tumor suppressor P53 and has been shown to be involved in P53-mediated apoptosis. Multiple alternatively spliced transcript variants, which encode different isoforms, have been reported for this gene. (provided by RefSeq)

Immunogen

BAX (AAH14175, 1 a.a. ~ 192 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDGSGEQPRGGGPTSSEQIMKTGALLLQGFIQDRAGRMGGEAPELALDPVPQDASTKKLSECLKRIGDELDSNMELQRMIAAVDTDSPREVFFRVAADMFSDGNFNWGRVVALFYFASKLVLKALCTKVPELIRTIMGWTLDFLRERLLGWIQDQGGWDGLLSYFGTPTWQTATIFVAGVLTASLTIWKKMG

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander Chota et al.
Frontiers in pharmacology, 13, 892490-892490 (2022-05-14)
Breast cancer is one of the most common types of cancer in women, and it is regarded as the second leading cause of cancer-related deaths worldwide. The present study investigated phytochemical profiling, in vitro anticancer effects of Dicoma anomala methanol
Alexander Chota et al.
Cells, 11(20) (2022-10-28)
Globally, lung cancer has remained the leading cause of morbidity and mortality in men and women. To enhance photodynamic therapeutic effects in vitro, the present study was designed to reduce dose-dependence in photodynamic therapy (PDT) and evaluate the anticancer effects
Anna Tankiewicz-Kwedlo et al.
International journal of molecular sciences, 19(4) (2018-04-25)
The Bruton’s tyrosine kinase (BTK) inhibitor LFM-A13 has been widely employed as an antileukemic agent, but applications in solid cancer have been found recently. The compound promotes apoptosis, has an antiproliferative effect, and increases cancer cell sensitivity to chemotherapy drugs.
Zhiyong Zhang et al.
Environmental toxicology, 36(8), 1557-1566 (2021-05-07)
Delphinidin is an anthocyanin that belongs to the group of flavonoids that exert numerous biological activities. However, the molecular mechanisms underlying the anticancer effects of delphinidin remain poorly understood. In our study we analyzed delphinidin modulate STAT-3 and MAPKinase signaling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service