Skip to Content
Merck
All Photos(1)

Key Documents

219555

Sigma-Aldrich

Cathepsin K Substrate II, Fluorogenic

An internally quenched, highly cathepsin K-selective, fluorogenic peptide substrate that is efficiently cleaved by CatK (kcat/Km = 0.426 µM⁻¹ S⁻¹), but not by Cathepsins B/L/S/F/H/V.

Synonym(s):

Cathepsin K Substrate II, Fluorogenic

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€157.00

€157.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€157.00

About This Item

Empirical Formula (Hill Notation):
C40H50N14O12
Molecular Weight:
918.91
UNSPSC Code:
51111800
NACRES:
NA.77

€157.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

bright yellow

solubility

DMSO: 5 mg/mL

fluorescence

λex 320 nm; λem 420 nm

shipped in

wet ice

storage temp.

−20°C

General description

An internally quenched, highly cathepsin K-selective, fluorogenic peptide substrate that is efficiently cleaved by CatK (kcat/Km = 0.426 µM-1 S-1), but not by Cathepsins B/L/S/F/H/V. Studies using WT and CatK-/- mice reveal that the substrate is not cleaved by other proteolytic enzymes present in fibroblast lysates. Emission Max: 420 NM, Excitation Max: 320 nM.

An internally quenched, highly cathepsin K-selective, fluorogenic peptide substrate that is efficiently cleaved by CatK (kcat/Km = 0.426 µM⁻¹ S⁻¹), but not by Cathepsins B/L/S/F/H/V.

Packaging

Packaged under inert gas

Warning

Toxicity: Standard Handling (A)

Sequence

Abz-His-Pro-Gly~Gly-Pro-Gln-EDDnp

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Lecaille, F., et al. 2003. Biochem. J.375, 307.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service