Skip to Content
Merck

Skip To

219482

Caveolin-1 Scaffolding Domain Peptide, Cell-permeable

Caveolin-1 scaffolding domain peptide (C1-SD82-101) fused at the N-terminus to the cell-permeable Antennapedia internalization sequence (43-58).

Synonym(s):

Caveolin-1 Scaffolding Domain Peptide, Cell-permeable, AP-Cav, Pen-C1-SD, RQIKIWFQNRRMKWKK-DGIWKASFTTFTVTKYWFYR, Cavtratin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Check Cart for Availability
€ 299,00

About This Item

Empirical Formula (Hill Notation):
C228H335N61O49S
Molecular Weight:
4746.54
NACRES:
NA.77
UNSPSC Code:
12352202
Assay:
≥95% (HPLC)
Form:
lyophilized solid
Storage condition:
OK to freeze, desiccated (hygroscopic), protect from light

€ 299,00


Check Cart for Availability

Request a Bulk Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

color

white

solubility

DMSO: 2 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

Caveolin-1 scaffolding domain peptide (C1-SD82-101) fused at the N-terminus to the cell-permeable Antennapedia internalization sequence (43-58). This peptide is reported to block eNOS activity and cellular NO release in vitro and reduce inflammation and tumorigenesis in vivo. Caveolin-1 interacts with several lipid-modified signaling ligands, such as EGFR, eNOS, G-protein α-subunits, PKCα, H-Ras, and Src, via the C1-SD82-101 sequence.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
eNOS
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Physical form

Supplied as a trifluoroacetate salt.

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 3 months at -20°C.

Other Notes

Bernatchez, P.N., et al. 2005. Proc. Natl. Acad. Sci. USA102, 761.
Williams, T.M., et al. 2004. J. Biol. Chem.279, 51630.
Gratton, J.P., et al. 2003. Cancer Cell4, 31.
Sukumaran, S.K., et al. 2002. J. Biol. Chem.277, 50716.
Liu, J., et al. 2002. J. Biol. Chem.277, 10661.
Bucci, M., et al. 2000. Nat. Med.6, 1362.
Okamoto, T., et al. 1998. J. Biol. Chem.273, 5419.
H-Arg-Gln-Ile-Lys-Ile-Trp-Phe-Gln-Asn-Arg-Arg-Met-Lys-Trp-Lys-Lys-Asp-Gly-Ile-Trp-Lys-Ala-Ser-Phe-Thr-Thr-Phe-Thr-Val-Thr-Lys-Tyr-Trp-Phe-Tyr-Arg-OH

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Toxicity: Carcinogenic / Teratogenic (D)

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
207001573096480026
assay

≥95% (HPLC)

assay

≥98% (HPLC)

assay

≥95% (HPLC)

assay

≥95% (HPLC)

form

lyophilized solid

form

lyophilized solid

form

lyophilized solid

form

solid

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

manufacturer/tradename

Calbiochem®

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage condition

OK to freeze, protect from light, desiccated (hygroscopic)

storage condition

OK to freeze, desiccated (hygroscopic), protect from light

storage condition

OK to freeze, desiccated (hygroscopic)

storage condition

OK to freeze, desiccated (hygroscopic), protect from light


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters