Skip to Content
Merck
All Photos(1)

Key Documents

207001

Sigma-Aldrich

Calcineurin Autoinhibitory Peptide, Cell-permeable

The Calcineurin Autoinhibitory Peptide, Cell-permeable controls the biological activity of Mn2+-stimulated calcineurin activity. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Calcineurin Autoinhibitory Peptide, Cell-permeable, 11R-CaN-AID, Ac-RRRRRRRRRRRGGGRMAPPRRDAMPSDA-NH₂

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€274.00

€274.00


Estimated to ship on28 May 2025



Select a Size

Change View
1 MG
€274.00

About This Item

Empirical Formula (Hill Notation):
C137H251N71O34S2
Molecular Weight:
3501.04
UNSPSC Code:
12352202
NACRES:
NA.77

€274.00


Estimated to ship on28 May 2025


Quality Level

Assay

≥98% (HPLC)

form

lyophilized solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

water: 1 mg/mL

shipped in

wet ice

storage temp.

−20°C

General description

A cell-permeable peptide that consists of the calcineurin (CaN) autoinhibitory domain (AID) fused to a poly-arginine-based protein transduction doamin (11R) and acts as an inhibitor of CaN phosphatase activity. Reported to block CaN-dependent NFAT nuclear translocation and NFAT promoter activity in neurons. Also reported to protect neuronal cells from glutamate-induced cell death.
A cell-permeable peptide that is composed of the calcineurin (CaN) autoinhibitory domain (AID) fused to a poly-arginine-based protein transduction domain (11R) and acts as an inhibitor of CaN phosphatase activity. Blocks CaN-dependent NFAT nuclear translocation and NFAT promoter activity in neurons. Protects neuronal cells from glutamate-induced cell death.

Biochem/physiol Actions

Cell permeable: yes
Primary Target
Blocks CaN-dependent NFAT nuclear translocation
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Carcinogenic / Teratogenic (D)

Sequence

Ac-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Arg-Gly-Gly-Gly-Arg-Met-Ala-Pro-Pro-Arg-Arg-Asp-Ala-Met-Pro-Ser-Asp-Ala-NH₂

Other Notes

Terada, H., et al. 2003. J. Neurochem.87, 1145.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service