Skip to Content
Merck
All Photos(1)

Key Documents

218785

Sigma-Aldrich

Caspase-10, Human, Recombinant, E. coli

Caspase-10, Human, Recombinant is expressed in E. coli as a single polypeptide. Fused at the N- and C- termini to a His•Tag sequence. Undergoes spontaneous autoprocessing to yield the active enzyme.

Synonym(s):

Mch4, FLICE-2

Sign Into View Organizational & Contract Pricing

Select a Size

3000 U
€726.00

€726.00


Estimated to ship on20 May 2025



Select a Size

Change View
3000 U
€726.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€726.00


Estimated to ship on20 May 2025


recombinant

expressed in E. coli

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

specific activity

≥150,000 units/mg protein
≥50,000 units/mL

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

M.W. 11,000 and 18,000 (heterodimer).
Recombinant, human caspase-10 fused at the N- and C- termini to a His•Tag sequence and expressed in E. coli as a single polypeptide. Subsequently undergoes spontaneous autoprocessing to yield the active enzyme. Useful for the study of enzyme regulation and kinetics, cleavage of target substrates, and inhibitor screening. MW: 11000 and 18000.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme that will cleave 1.0 pmol Ac-IETD-pNA (Cat. No. 368057) per min at 25°C, pH 7.4.

Physical form

In 100 mM NaCl, 50 mM HEPES, 10 mM DTT, 1 mM EDTA, 10% glycerol, 0.1% CHAPS, 0.09% APO-10, pH 7.4.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Kang, J.J., et al. 1999. J. Biol. Chem. 274, 3189.
Ng, P.W., et al. 1999. J. Biol. Chem. 274, 10301.
Vincenz, C., and Dixit, V.M. 1997. J. Biol. Chem. 272, 6578.
Srinivasula, S.M., et al. 1996. Proc. Natl. Acad. Sci. USA 93, 14486.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service