Skip to Content
Merck
All Photos(1)

Key Documents

218765

Sigma-Aldrich

Caspase-9 Substrate I, Fluorogenic

A fluorogenic substrate for caspase-9 (ICE-LAP6, Mch6).

Synonym(s):

Caspase-9 Substrate I, Fluorogenic, Ac-LEHD-AFC

Sign Into View Organizational & Contract Pricing

Select a Size

1 MG
€109.00
5 MG
€403.00

€109.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
1 MG
€109.00
5 MG
€403.00

About This Item

Empirical Formula (Hill Notation):
C33H38F3N7O11
CAS Number:
Molecular Weight:
765.69
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€109.00


Estimated to ship on21 May 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated
protect from light

color

white

solubility

DMSO: 5 mg/mL

fluorescence

λex ~400 nm
λem ~505 nm

shipped in

ambient

storage temp.

−20°C

SMILES string

FC(F)(F)C1=CC(=O)Oc2c1ccc(c2)NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)[C@@H](NC(=O)C)CC(C)C)CCC(=O)O)Cc3[nH]cnc3)CC(=O)O

InChI

1S/C33H38F3N7O11/c1-15(2)8-22(39-16(3)44)31(52)41-21(6-7-26(45)46)29(50)42-23(9-18-13-37-14-38-18)32(53)43-24(12-27(47)48)30(51)40-17-4-5-19-20(33(34,35)36)11-28(49)54-25(19)10-17/h4-5,10-11,13-15,21-24H,6-9,12H2,1-3H3,(H,37,38)(H,39,44)(H,40,51)(H,41,52)

InChI key

HULKIXRFKRCRHD-ZJZGAYNASA-N

General description

A fluorogenic substrate for caspase-9 (ICE-LAP6, Mch6). May also act as a substrate for caspase-4 and caspase-5. Reaction can be monitored visually or quantitatively by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore.

Biochem/physiol Actions

Cell permeable: no
Primary Target
A fluorogenic substrate for caspase-9
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Ac-Leu-Glu-His-Asp-AFC

Physical form

Supplied as a trifluoroacetate salt.

Reconstitution

Following reconstitution aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Ito, A., et al. 1999. Biochim. Biophys. Acta 1452, 263.
Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Thornberry, N.A., et al. 1997. J. Biol. Chem. 272, 17907.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service