Skip to Content
Merck
All Photos(2)

Key Documents

418773

Sigma-Aldrich

3,4-Dihydro-3-methyl-2(1H)-quinazolinone

99%

Sign Into View Organizational & Contract Pricing

Select a Size

50 G
€58.80

€58.80


Estimated to ship on14 April 2025


Request a Bulk Order

Select a Size

Change View
50 G
€58.80

About This Item

Empirical Formula (Hill Notation):
C9H10N2O
CAS Number:
Molecular Weight:
162.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€58.80


Estimated to ship on14 April 2025


Request a Bulk Order

Quality Level

Assay

99%

mp

200-203 °C (lit.)

SMILES string

CN1Cc2ccccc2NC1=O

InChI

1S/C9H10N2O/c1-11-6-7-4-2-3-5-8(7)10-9(11)12/h2-5H,6H2,1H3,(H,10,12)

InChI key

GINUUQLRYXHAPB-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy P C Rooney et al.
Angewandte Chemie (International ed. in English), 53(24), 6126-6130 (2014-05-14)
The benzoxazinone and dihydroquinoxalinone fragments were employed as novel acetyl lysine mimics in the development of CREBBP bromodomain ligands. While the benzoxazinone series showed low affinity for the CREBBP bromodomain, expansion of the dihydroquinoxalinone series resulted in the first potent

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service