Skip to Content
Merck
All Photos(1)

Documents

218746

Sigma-Aldrich

Caspase-1 Inhibitor VI

≥95% (HPLC), powder, Caspase-1 inhibitor, Calbiochem®

Synonym(s):

Caspase-1 Inhibitor VI, Z-YVAD-FMK

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C31H39FN4O9
Molecular Weight:
630.66
UNSPSC Code:
12352200
NACRES:
NA.77

product name

Caspase-1 Inhibitor VI, Caspase-1 Inhibitor VI, Z-YVAD-FMK, is a potent, cell-permeable, and irreversible inhibitor of caspase-1, caspase-4, and caspase-5.

Quality Level

Assay

≥95% (HPLC)
≥95% (TLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated

color

white

solubility

DMSO: 5 mg/mL

shipped in

ambient

storage temp.

−20°C

General description

A potent, cell-permeable and irreversible inhibitor of caspase-1 (ICE).
A potent, cell-permeable, and irreversible inhibitor of caspase-1 (ICE), caspase-4, and caspase-5.

Biochem/physiol Actions

Cell permeable: yes
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Standard Handling (A)

Sequence

Z-Tyr-Val-Ala-Asp(OMe)-CH₂F*

Other Notes

Thornberry, N.A., and Lazebnik, Y. 1998. Science 281, 1312.
Eldadah, B.A., et al. 1997. J. Neurosci. 17, 6105.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Priyoshi Lahiri et al.
Infection and immunity, 92(2), e0034223-e0034223 (2024-01-08)
Digital dermatitis (DD) is a skin disease in cattle characterized by painful inflammatory ulcerative lesions in the feet, mostly associated with local colonization by Treponema spp., including Treponema phagedenis. The reason why most DD lesions remain actively inflamed and progress
Bo Shi et al.
World journal of pediatrics : WJP (2023-06-30)
Macrophages are involved in various immune inflammatory disease conditions. This study aimed to investigate the role and mechanism of macrophages in regulating acute intestinal injury in neonatal necrotizing enterocolitis (NEC). CD68, nucleotide-binding oligomerization domain, leucine-rich repeat, and pyrin domain-containing 3
Inflammatory macrophage to hepatocyte signals can be prevented by extracellular vesicle reprogramming.
Ghosh, et al.
Journal of Cell Science, 136 (2023)
Ye Eun Jeong et al.
Annals of clinical and translational neurology, 10(2), 246-265 (2022-12-28)
Neurodegeneration induced by inflammatory stress in multiple sclerosis (MS) leads to long-term neurological disabilities that are not amenable to current immunomodulatory therapies. Here, we report that neuronal downregulation of Splicing factor 3b subunit 2 (SF3B2), a component of U2 small
Elizabeth M Gordon et al.
The Journal of allergy and clinical immunology, 144(2), 426-441 (2019-03-16)
House dust mite (HDM)-challenged Apoe-/- mice display enhanced airway hyperreactivity and mucous cell metaplasia. We sought to characterize the pathways that induce apolipoprotein E (APOE) expression by bronchoalveolar lavage fluid (BALF) macrophages from asthmatic subjects and identify how APOE regulates

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service