Skip to Content
Merck
All Photos(1)

Key Documents

1.99012

Supelco

Buffer Solution

(citric acid, sodium hydroxide, hydrogen chloride) tracable to SRM from NIST and PTB pH 2.00 (25°C) Certipur®

Synonym(s):

Buffer solution pH 2.0 (citric acid/sodium hydroxide/hydrogen chloride)

Sign Into View Organizational & Contract Pricing

Select a Size

30 X 30 ML
€101.00

€101.00


Estimated to ship on21 May 2025


Request a Bulk Order

Select a Size

Change View
30 X 30 ML
€101.00

About This Item

UNSPSC Code:
12161704
NACRES:
NA.25

€101.00


Estimated to ship on21 May 2025


Request a Bulk Order

grade

certified reference material

Quality Level

product line

Certipur®

pH

2.0 (25 °C in H2O)

density

1.00 g/cm3 at 20 °C

application(s)

environmental
food and beverages
general analytical
industrial qc
pharmaceutical

format

mixture

storage temp.

15-25°C

General description

It is a certified buffer solution of citric acid/sodium hydroxide/hydrogen chloride, gravimetrically prepared using high-quality reagents at a registered calibration laboratory. It has a pH value of 2.00 at 25°C, measured using a combined glass electrode following a 5-point calibration method. The measurement is made in accordance with DIN 19268 using reference buffer solutions compliant to DIN 19266, NIST, IUPAC, Ph. Eur, and USP. The pH value of the buffer solution depends on the temperature and therefore a constant temperature must be maintained during its use.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at http://www.sigma-aldrich.com to view certified values, including uncertainty and expiry date.

Application

The buffer solution is used as a standard in analytical procedures for the calibration of pH measuring instruments or pH electrodes and as a control for the measurement of pH of other solutions.

Features and Benefits

  • Measured and qualified at an ISO/IEC 17025 and ISO 17034 accredited laboratory
  • Directly traceable to secondary reference standards from NIST and primary certified reference materials from PTB
  • Ready-to-use solution available in 30 mL sachets
  • Accompanied with a detailed and batch-specific certificate of analysis (CoA) for an effective quality management system
  • Product label follows the ISO Guide 31 and contains information about ordering number, article description, lot number, safety data, and expiry date
  • Packaging material subjected to comprehensive stability studies as per ISO Guide 35
  • An uncertainty of +/- 0.02 in the pH value at 25°C

Legal Information

CERTIPUR is a registered trademark of Merck KGaA, Darmstadt, Germany

recommended

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service