Skip to Content
Merck
All Photos(1)

Key Documents

171257

Millipore

Ampicillin, Sodium Salt, Sterile-Filtered Aqueous Solution, Cell Culture Tested

sterile; sterile-filtered, suitable for cell culture

Synonym(s):

Ampicillin, Sodium Salt, Sterile-Filtered Aqueous Solution, Cell Culture Tested, 6-[D(–) α-Aminophenylacetamido]penicillanic Acid, Na

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H18N3O4S · Na
CAS Number:
Molecular Weight:
371.39
MDL number:
UNSPSC Code:
12352200

sterility

sterile; sterile-filtered

Quality Level

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze

technique(s)

cell culture | mammalian: suitable

impurities

≤2 EU/mg Endotoxins (dry weight)

shipped in

wet ice

storage temp.

−20°C

InChI

1S/C16H19N3O4S.Na/c1-16(2)11(15(22)23)19-13(21)10(14(19)24-16)18-12(20)9(17)8-6-4-3-5-7-8;/h3-7,9-11,14H,17H2,1-2H3,(H,18,20)(H,22,23);/q;+1/p-1/t9-,10-,11+,14-;/m1./s1

InChI key

KLOHDWPABZXLGI-YWUHCJSESA-M

General description

A β-lactam antibiotic that inhibits bacterial cell-wall synthesis. Active against Gram-negative bacteria. Inactivated by lactamases. Suitable for use in research that uses ampicillin-resistant plasmids.

Warning

Toxicity: Harmful (C)

Physical form

Supplied at 100 mg/ml in H₂O.

Reconstitution

Following initital thaw, aliquot and freeze (-20°).

Other Notes

Nakajima, M., et al. 1994. Neurosci. Lett.176, 161.
Wu, M.H., and Yung, B.Y. 1994. Eur. J. Pharmacol.270, 203.
Betzel, C., et al. 1993. Biochim. Biophys. Acta1161, 47.
Yung, B.Y., et al. 1992. Int. J. Cancer52, 317.
Martin, S.J., et al. 1990. J. Immunol.145, 1859.
Yung, B.Y., et al. 1990. Cancer Res.50, 5987.
White, R.J., and Phillips, D.R. 1985. Biochemistry27, 9122.
Madharavao, M., et al. 1978. J. Med. Chem.21, 958.
Sengupta, S.K., et al. 1975. J. Med. Chem.18, 1175.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service