Skip to Content
Merck
All Photos(1)

Key Documents

171256

Sigma-Aldrich

AMPK Activator

The AMPK Activator, also referenced under CAS 849727-81-7, modulates the biological activity of AMPK. This small molecule/inhibitor is primarily used for Activators/Inducers applications.

Synonym(s):

AMPK Activator, 5-(3-(4-(2-(4-Fluorophenyl)ethoxy)phenyl)propyl)furan-2-carboxylic acid, D942, AMPK Signaling Activator I

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€196.00

€196.00


Estimated to ship on02 June 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€196.00

About This Item

Empirical Formula (Hill Notation):
C22H21FO4
CAS Number:
Molecular Weight:
368.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

€196.00


Estimated to ship on02 June 2025


Request a Bulk Order

Quality Level

Assay

≥95% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

light yellow

solubility

DMSO: 10 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)CCOc2ccc(cc2)CCCc3[o]c(cc3)C(=O)O

InChI

1S/C22H21FO4/c23-18-8-4-17(5-9-18)14-15-26-19-10-6-16(7-11-19)2-1-3-20-12-13-21(27-20)22(24)25/h4-13H,1-3,14-15H2,(H,24,25)

InChI key

MPLLLQUZNJSVTK-UHFFFAOYSA-N

General description

A cell-permeable furancarboxylic acid compound that activates celular AMP-activated protein kinase (AMPK) activity by inhibiting mitochondrial complex I, and thus elevating intracellular AMP level. Shown to enhance glucose uptake in L6 myocytes (EC50 = 11.7 µM). Reduces blood glucose levels in ZDF rats in vivo (oral dose 30-100 mg/kg). Displays negligible cytotoxicity at pharmacological doses.

Biochem/physiol Actions

Cell permeable: yes
EC50 = 11.7 µM enhancing glucose uptake in L6 myocytes
Primary Target
AMPK
Product does not compete with ATP.
Reversible: no

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Kosaka, T., et al. 2005. Anal. Chem.77, 2050.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service