Skip to Content
Merck
All Photos(1)

Documents

171261

Sigma-Aldrich

Dorsomorphin

≥95% (HPLC), liquid, AMPK inhibitor, Calbiochem

Synonym(s):

InSolution AMPK Inhibitor, Compound C, Dorsomorphin, AMPK Inhibitor I, BMP Inhibitor I

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H25N5O
CAS Number:
Molecular Weight:
399.49
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

product name

AMPK Inhibitor, Compound C, InSolution, ≥95%, 10 mM, AMPK Inhibitor

Quality Level

Assay

≥95% (HPLC)

form

liquid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

shipped in

wet ice

storage temp.

2-8°C

InChI

1S/C24H25N5O/c1-2-12-28(13-3-1)14-15-30-22-6-4-19(5-7-22)21-16-26-24-23(17-27-29(24)18-21)20-8-10-25-11-9-20/h4-11,16-18H,1-3,12-15H2

InChI key

XHBVYDAKJHETMP-UHFFFAOYSA-N

General description

A cell-permeable pyrrazolopyrimidine compound that inhibits against KDR/VEGFR2, ALK2/BMPR-I, AMPK kinase activity (IC50 = 25.1, 148, and 234.6 nM, respectively), while exhibiting much reduced or little effect toward ALK5/TGFβR-I, ZAPK, Syk, PKCθ, PKA, or JAK3. Shown to block both BMP pathway-dependent dorsoventral development (EC100 = 2.5 µM) and VEGF signaling-dependent intersomitic vessel formation (EC50 = 5 µM) in zebrafish embryo in vivo. Commonly used in combination with AMPK activators AICAR (Cat. No. 123040) and/or Metformin (Cat. No. 317240) for studying AMPK-dependent cellular events in vitro and physiological responses in animals in vivo.

Biochem/physiol Actions

Cell permeable: yes
EC100 = 2.5 µM against BMP pathway-dependent dorsoventral development; EC50 = 5 µM against VEGF signaling-dependent intersomitic vessel formation in zebrafish embryo in vivo
Primary Target
AMPK
Product competes with ATP.
Reversible: yes
Target IC50: 25.1, 148, and 234.6 nM, against KDR/VEGFR2, ALK2/BMPR-I, AMPK kinase activity, respectively

Packaging

Packaged under inert gas

Warning

Toxicity: Irritant (B)

Physical form

Supplied as a 10 mM (1 mg/250 µl) solution of AMPK Inhibitor, Compound C (Cat. No. 171260) in DMSO.

Reconstitution

Following initial use, aliquot and freeze (-20°C). Once opened, stock solutions are stable for up to 3 months at -20°C.

Other Notes

Hao, J., et al. 2010. ACS Chem.Biol.5, 245.
Kim, E.K., et al. 2004. J. Biol. Chem.279, 19970.
Lee, M., et al. 2003. J. Biol. Chem.278, 39653.
Inoki, K., et al. 2003. Cell115, 577.
Fryer, L.G., 2002. FEBS Lett.531, 189.
Zhou, G., et al. 2001. J. Clin. Invest.108, 1167.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

188.6 °F - closed cup - (Dimethylsulfoxide)

Flash Point(C)

87 °C - closed cup - (Dimethylsulfoxide)


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao-Yao Chang et al.
Frontiers in immunology, 13, 820524-820524 (2022-03-01)
P2Y1 receptor is a G-protein-coupled receptor that plays a critical role in the immune response of inflammatory bowel diseases. However, its regulatory effects on CD4+ T cell response have not been fully elucidated. The study aimed to characterize the role
Ernesto Pena Calderin et al.
Molecular metabolism, 66, 101637-101637 (2022-11-19)
Physical activity has been shown to reduce the risk of CVD mortality in large-cohort longitudinal studies; however, the mechanisms underpinning the beneficial effects of exercise remain incompletely understood. Emerging data suggest that the risk reducing effect of exercise extends beyond
Reetta A E Eriksson et al.
Frontiers in medicine, 9, 1052318-1052318 (2022-12-31)
Gene therapy would greatly benefit from a method to regulate therapeutic gene expression temporally. Riboswitches are small RNA elements that have been studied for their potential use in turning transgene expression on or off by ligand binding. We compared several
Yidi Wang et al.
Cells, 11(19) (2022-10-15)
Crouzon syndrome ([OMIM] #123500) caused by FGFR2 mutation is an autosomal dominant syndrome with craniosynostosis, the underlying mechanism of which remains obscure. First, whole exome sequencing was used to screen the possible pathogenic variant in two sporadic patients with Crouzon
Karina Krotova et al.
Human gene therapy, 31(19-20), 1124-1131 (2020-06-05)
Adeno-associated virus (AAV)-based gene therapy is undergoing major expansion into clinical practice, with two treatments currently being granted Food and Drug Administration (FDA) approval. However, the presence of pre-existing neutralizing antibodies (NAB) is one of the significant hurdles for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service