Skip to Content
Merck
All Photos(1)

Key Documents

165304

Sigma-Aldrich

D-(–)-2-Amino-5-phosphonopentanoic Acid

≥97% (HPLC), solid, NMDA receptor antagonist, Calbiochem®

Synonym(s):

D-(–)-2-Amino-5-phosphonopentanoic Acid, D-AP5, NMDA Antagonist II, APV, D-APV, 2-APV, D-2-amino-5-phosphonovalerate

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
€199.00

€199.00


Estimated to ship on29 May 2025


Request a Bulk Order

Select a Size

Change View
5 MG
€199.00

About This Item

Empirical Formula (Hill Notation):
C5H12NO5P
CAS Number:
Molecular Weight:
197.13
MDL number:
UNSPSC Code:
12352106
NACRES:
NA.77

€199.00


Estimated to ship on29 May 2025


Request a Bulk Order

Product Name

D-(–)-2-Amino-5-phosphonopentanoic Acid, Active enantiomer of DL-2-amino-5-phosphonopentanoic acid (AP5) that is a commonly used as a competitive NMDA receptor antagonist.

Quality Level

Assay

≥97% (HPLC)

form

solid

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
desiccated (hygroscopic)
protect from light

color

white

solubility

dilute aqueous base: 1 mg/mL
water: 1 mg/mL

shipped in

ambient

storage temp.

2-8°C

SMILES string

[P](=O)([O-])([O-])CCC[C@@H]([N+H3])C(=O)[O-]

InChI

1S/C5H12NO5P/c6-4(5(7)8)2-1-3-12(9,10)11/h4H,1-3,6H2,(H,7,8)(H2,9,10,11)/p-2/t4-/m1/s1

InChI key

VOROEQBFPPIACJ-SCSAIBSYSA-L

General description

Active enantiomer of DL-2-amino-5-phosphonopentanoic acid (AP5) that is a commonly used as a competitive NMDA receptor antagonist. Also inhibits the capsaicin-induced release of substance P from spinal cord.

Biochem/physiol Actions

Cell permeable: no
Primary Target
NMDA receptor antagonist
Product does not compete with ATP.
Reversible: no

Warning

Toxicity: Irritant (B)

Reconstitution

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Other Notes

Malcangio, M., et al. 1998. Br. J. Pharmacol. 125, 1625.
Schulte, M.K., et al. 1994. Brain Res. 649, 203.
Davis, S., et al. 1992. J. Neurosci. 12, 21.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Jing Zhou et al.
eLife, 10 (2021-03-05)
Callosal projections from primary somatosensory cortex (S1) are key for processing somatosensory inputs and integrating sensory-motor information. How the callosal innervation pattern in S1 is formed during early postnatal development is not clear. We found that the normal termination pattern
Yu-Hong Zhang et al.
Cell stem cell, 28(8), 1483-1499 (2021-04-23)
The hypothalamus contains an astounding heterogeneity of neurons that regulate endocrine, autonomic, and behavioral functions. However, its molecular developmental trajectory and origin of neuronal diversity remain unclear. Here, we profile the transcriptome of 43,261 cells derived from Rax+ hypothalamic neuroepithelium
Hitomi Abe et al.
iScience, 25(3), 103927-103927 (2022-03-15)
Understanding the molecular mechanisms of gene regulation is pivotal for understanding how cells establish and modify their identities and functions. Multiple transcription factors (TFs) coordinate to alter gene expression in cells; however, a method to quantitatively analyze the activity of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service