Skip to Content
Merck
All Photos(1)

Key Documents

1.50974

Supelco

Superspher® 60 RP-select B (4 µm) HPLC Columns

L x I.D. 75 mm x 4 mm, LiChroCART HPLC cartridge, manuCART holder required for use

Synonym(s):

TM=["Superspher"] C8 TM=["LICHROCART"] cartridge for HPLC

Sign Into View Organizational & Contract Pricing

Select a Size

3 UNITS
€1,060.00

€1,060.00


Estimated to ship on25 May 2025



Select a Size

Change View
3 UNITS
€1,060.00

About This Item

UNSPSC Code:
41115709
NACRES:
SB.52

€1,060.00


Estimated to ship on25 May 2025


Product Name

Superspher® 60 RP-select B LiChroCART 75-4, suitable for HPLC

material

stainless steel column

Quality Level

Agency

suitable for USP L7

product line

Superspher®

feature

endcapped: no

parameter

250 bar pressure
60 °C max. temp.

technique(s)

HPLC: suitable
mass spectrometry (MS): suitable (Low - Medium)

column L × I.D.

7.5 cm × 4 mm

matrix

fully porous particle

matrix active group

C8 bonding phase

particle size

4 μm

pore size

60 Å pore size

operating pH range

2-7.5

separation technique

reversed phase

storage temp.

no temp limit

General description

HPLC cartridge
Superspher® RP-select B is a highly reproducible traditionally produced spherical silica gel carriers with reversed phase properties and the hydrophobicity of a C-8 phase. This carrier is very well suited for the separation of basic compounds, but with still good properties for the determination of neutral and acidic compounds. The high performance silica carrier with a particle size of 4µm yields an excellent pressure/separation performance ratio.This sorbent is packed into a LiChroCART 75-4 HPLC cartridge. The connection to the HPLC system is being accomplished by use of the manu-CART NT cartrige holder (1.51486).

Application


  • Screening, library-assisted identification and validated quantification of 23 benzodiazepines, flumazenil, zaleplone, zolpidem and zopiclone in plasma: This study demonstrates the application of liquid chromatography/mass spectrometry with atmospheric pressure chemical ionization for the screening and quantification of various compounds in plasma, utilizing the high-resolution capabilities of the Superspher 60 RP-select B column (Kratzsch et al., 2004).

  • Screening, library-assisted identification and validated quantification of fifteen neuroleptics and three of their metabolites in plasma: Employing liquid chromatography/mass spectrometry with atmospheric pressure chemical ionization, this study utilized the Superspher 60 RP-select B to detect and quantify neuroleptics in plasma, showcasing its utility in pharmaceutical and clinical research (Kratzsch et al., 2003).

  • Validated assay for quantification of oxcarbazepine and its active dihydro metabolite 10-hydroxycarbazepine in plasma: This research highlights the column′s application in therapeutic drug monitoring using atmospheric pressure chemical ionization liquid chromatography/mass spectrometry, reflecting its precision and reliability in clinical pharmacology (Maurer et al., 2002).

Analysis Note

Pressure: ≤ 50 bar
Theoretical Plates (N/m) (1,2-Benzanthracene): ≥ 85000
Symmetry (1,2-Benzanthracene): 0.8 - 1.4
Capacity factor (1,2-Benzanthracene): ≥ 2.5

Legal Information

LICHROCART is a trademark of Merck KGaA, Darmstadt, Germany
Superspher is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service