Skip to Content
Merck
All Photos(1)

Key Documents

14-785-M

Sigma-Aldrich

PI3 Kinase (p110α/p85α) murine, 10 µg

Complex of N-terminal 6His-tagged, recombinant, full-length mouse p110α & untagged, recombinant, full length mouse p85α. Coexpressed by baculovirus in Sf21 insect cells. For use in Kinase Assays.

Synonym(s):

PI3K Enzyme

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€581.00

€581.00


Estimated to ship on21 May 2025



Select a Size

Change View
10 μG
€581.00

About This Item

UNSPSC Code:
12352200
eCl@ss:
32160405
NACRES:
NA.32

€581.00


Estimated to ship on21 May 2025


biological source

mouse

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 129 kDa (p110α)
Mw 83.6 kDa (p85α)

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

mouse ... PIK3CA(18706)

General description

Complex of N-terminal 6His-tagged, recombinant, full-length mouse p110αand untagged, recombinant, full length mouse p85α. Coexpressed by baculovirus in Sf21 insect cells.
Product Source: Coexpressed by baculovirus in Sf21 insect cells.

Application

Research Category
Inflammation & Immunology

Biochem/physiol Actions

Protein Target: PI3Kα/p85α
Target Sub-Family: PI3K/PI4K

Packaging

Also available in 250μg size --call for pricing and availability and reference catalog number 14-785M when ordering the 250μg size.

Physical form

Ni2+/NTA-agarose

Storage and Stability

Store at -70°C from date of shipment. For maximum recovery of product, centrifuge original vial prior to removing the cap.

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service