Skip to Content
Merck
All Photos(1)

Key Documents

412295

Sigma-Aldrich

4-(Trifluoromethyl)phenylhydrazine

96%

Synonym(s):

1-(4-Trifluoromethylphenyl)hydrazine, 4-Trifluoromethylphenylhydrazine, [4-(Trifluoromethyl)phenyl]hydrazine, p-Trifluoromethylphenylhydrazine

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
€78.50

€78.50


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
€78.50

About This Item

Linear Formula:
CF3C6H4NHNH2
CAS Number:
Molecular Weight:
176.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

€78.50


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

bp

118-122 °C/17 mmHg (lit.)

mp

63-65 °C (lit.)

functional group

fluoro
hydrazine

storage temp.

2-8°C

SMILES string

NNc1ccc(cc1)C(F)(F)F

InChI

1S/C7H7F3N2/c8-7(9,10)5-1-3-6(12-11)4-2-5/h1-4,12H,11H2

InChI key

DBNLGTYGKCMLLR-UHFFFAOYSA-N

General description

4-(Trifluoromethyl)phenylhydrazine participates as phenylhydrazine-based reductant containing fluorine atoms in the one-step reduction and functionalization of graphene oxide.[1]

Application

4-(Trifluoromethyl)phenylhydrazine may be used for the synthesis of 2-iodo-9-trifluoromethyl-paullone.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Su-Hyeon Kim et al.
Nanoscale, 6(13), 7183-7187 (2014-05-08)
A one-step reduction and functionalization of graphene oxide (FrGO) was easily achieved using a novel phenylhydrazine-based reductant containing fluorine atoms, which can induce p-type doping due to its high electronegativity. The FrGO-based OPV exhibited a high power conversion efficiency of
C Kunick et al.
Bioorganic & medicinal chemistry letters, 10(6), 567-569 (2000-03-31)
9-Trifluoromethyl-paullones with a carbon chain in the 2-position were synthesized by palladium-catalyzed coupling reactions of a 2-iodoprecursor with terminal alkenes or alkynes, respectively. The introduction of a 2-cyanoethyl substituent led to a significant enhancement of CDK1/cyclin B inhibiting property and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service