Skip to Content
Merck
All Photos(1)

Key Documents

14-688-M

Sigma-Aldrich

PKG1α Protein, active, 10 µg

Active, N-terminal 6His-tagged, recombinant, full length PKG1α. For use in Kinase Assays.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 80 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

Gene Information

human ... PRKG1(5592)

General description

N-terminal 6His-tagged, recombinant, full length PKG1α.
Product Source: PKG1α, expressed by baculovirus in Sf21 cells.

Application

Research Category
Inflammation & Immunology

Biochem/physiol Actions

Protein Target: PKG1α
Target Sub-Family: AGC

Quality

routinely evaluated by phosphorylation of PAKtide (RRRLSFAEPG).

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C from date of shipment

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physiology and pathophysiology of vascular signaling controlled by guanosine 3',5'-cyclic monophosphate-dependent protein kinase.
Birschmann, Ingvild and Walter, Ulrich
Acta Biochimica Polonica, 51, 397-404 (2004)
Sequential activation of soluble guanylate cyclase, protein kinase G and cGMP-degrading phosphodiesterase is necessary for proper induction of long-term potentiation in CA1 of hippocampus. Alterations in hyperammonemia.
Monfort, Pilar, et al.
Neurochemistry International, 45, 895-901 (2004)
Mary G Johlfs et al.
PloS one, 10(7), e0132105-e0132105 (2015-07-02)
Profiling cellular proteome is critical to understanding signal integration during cell fate determination. In this study, the capability of capillary isoelectric focusing (cIEF) immunoassays to detect post-translational modifications (PTM) of protein isoforms is demonstrated. cIEF immunoassays exhibit protein detection sensitivity
Modulation of synaptic function by cGMP and cGMP-gated cation channels.
Barnstable, Colin J, et al.
Neurochemistry International, 45, 875-884 (2004)
Zhenyu Li et al.
Cell, 112(1), 77-86 (2003-01-16)
It is currently accepted that cGMP-dependent protein kinase (PKG) inhibits platelet activation. Here, we show that PKG plays an important stimulatory role in platelet activation. Expression of recombinant PKG in a reconstituted cell model enhanced von Willebrand factor (vWF)-induced activation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service