Skip to Content
Merck
All Photos(1)

Key Documents

74767

Supelco

Cyhalothrin

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
€61.70

€61.70


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
€61.70

About This Item

Empirical Formula (Hill Notation):
C23H19ClF3NO3
CAS Number:
Molecular Weight:
449.85
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

€61.70


Check Cart for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

PESTANAL®

Assay

≥98.0% (mixture of 4 isomers, HPLC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

mp

48-54 °C

format

neat

SMILES string

FC(F)(F)\C(=C\C1C(C1C(=O)OC(c2cc(ccc2)Oc3ccccc3)C#N)(C)C)\Cl

InChI

1S/C23H19ClF3NO3/c1-22(2)17(12-19(24)23(25,26)27)20(22)21(29)31-18(13-28)14-7-6-10-16(11-14)30-15-8-4-3-5-9-15/h3-12,17-18,20H,1-2H3/b19-12-

InChI key

ZXQYGBMAQZUVMI-UNOMPAQXSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 3

1 of 3

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service