Skip to Content
Merck
All Photos(1)

Documents

1.46093

Millipore

Chocolate Agar +LTH - SDV

90 mm ready-to-use plates for enumeration of total aerobic microbial count

Synonym(s):

Chocolate Blood agar, CHOC agar, CBA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106217
NACRES:
NA.74

material

polystyrene plate

Quality Level

Agency

non-compendial

sterility

sterile; γ-irradiated

form

plate (ready-to-use)

feature

2D bar code
ready-to-use

D

90 mm

pH

7.3 ( in H2O)

application(s)

air monitoring
microbiology
pharmaceutical

compatibility

for use with Settle Plate

storage temp.

2-8°C

suitability

bacteria
fungi

Related Categories

General description

Chocolate Agar + LTH – SDV is a ready-to-use universal blood-containing culture medium for enrichment and isolation of fastidious bacteria, yeast, and molds. This medium is suitable for the detection of aerobic as well as anaerobic microorganisms. It is characterized by a rich nutritive basis. Dextrose is a source of carbon and energy. In addition, the growth supplements haemin and NAD are released from the sheep blood erythrocytes during heating. These components provide good growth conditions for fastidious aerobic and anaerobic microorganisms. The medium is supplemented with neutralizers such as lecithin, polysorbate (Tween®) 80, and histidine to efficiently neutralize disinfectants.

Application

Chocolate Agar + LTH in 90 mm settle plates is designed for the enumeration of the total aerobic microbial count in air via active or passive air monitoring as well as fingerprints of personnel in Isolators and Clean Rooms.

Legal Information

TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

Settle plates for isolators and critical cleanrooms

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service