Skip to Content
Merck
All Photos(1)

Key Documents

14-526-M

Sigma-Aldrich

Met Protein, active, 10 0

Active, N-terminal His6-tagged recombinant human Met residues 974-end, for use in Kinase Assays.

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€581.00
250 μG
€3,780.00

€581.00


Estimated to ship on17 May 2025



Select a Size

Change View
10 μG
€581.00
250 μG
€3,780.00

About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.41

€581.00


Estimated to ship on17 May 2025


biological source

human

Quality Level

recombinant

expressed in baculovirus infected Sf21 cells

mol wt

Mw 50 kDa

manufacturer/tradename

Upstate®

technique(s)

activity assay: suitable (kinase)

NCBI accession no.

UniProt accession no.

General description

N-terminal His6-tagged recombinant human Met residues 974-end
Product Source: expressed by baculovirus in Sf21 insect cells

Application

Research Category
Metabolism

Inflammation & Immunology
Research Sub Category
Obesity

Metabolic Disorders

Osteoporosis

Arthritis

Biochem/physiol Actions

Protein Target: Met
Target Sub-Family: TK

Packaging

2 vials at 5μg each
Also available in 250μg size (2x125μg)--call for pricing and availability and reference catalog number 14-526M when ordering the 250μg size.

Quality

routinely evaluated by phosphorylation of KKKSPGEYVNIEFG

Physical form

Ni2+/NTA-agarose

Storage and Stability

6 months at -70°C

Other Notes

For Specific Activity data, refer to the Certificate of Analysis for individual lots of this enzyme.

Legal Information

UPSTATE is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service