Skip to Content
Merck
All Photos(1)

Key Documents

325885

Sigma-Aldrich

c-MET, GST-Fusion, Human, Recombinant, S. frugiperda

c-MET, GST-Fusion, Human, Recombinant (aa 956-1390) fused at the N-terminus to a GST-His6-thrombin cleavage site and expressed in S. frugiperda. c-Met is a receptor tyrosine kinase activated by HGF.

Synonym(s):

Hepatocyte growth factor receptor tyrosine kinase

Sign Into View Organizational & Contract Pricing

Select a Size

10 μG
€551.00

€551.00


Estimated to ship on16 April 2025



Select a Size

Change View
10 μG
€551.00

About This Item

UNSPSC Code:
12352202
NACRES:
NA.77

€551.00


Estimated to ship on16 April 2025


recombinant

expressed in baculovirus infected Sf21 cells

Quality Level

form

liquid

specific activity

≥30 units/mg protein

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

shipped in

wet ice

storage temp.

−70°C

General description

Recombinant, human MET (amino acids 956-1390) (Genebank entry NM_000245) fused at the N-terminus to a GST-His6-thrombin cleavage site sequence and expressed in S. frugiperda insect cells using a baculovirus expression system. c-Met is a receptor tyrosine kinase activated by hepatocyte growth factor (HGF)/scatter factor (SF). MET is a plasminogen-like protein thought to be a humoral mediator of liver regeneration.
Recombinant, human c-MET (amino acids 956-1390) (Genebank entry NM_000245) fused at the N-terminus to a GST-His6-thrombin cleavage site sequence and expressed in S. frugiperda insect cells using a baculovirus expression system. c-Met is a receptor tyrosine kinase activated by hepatocyte growth factor (HGF)/scatter factor (SF). c-MET is a plasminogen-like protein thought to be a humoral mediator of liver regeneration.

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Unit Definition

One unit is defined as the amount of enzyme required to transfer 1 nmol phosphate to Poly(Ala, Glu, Lys, Tyr)6:2:5:1 per min at 30°C, using variable concentrations of ATP (0.1-1.48 µM).

Physical form

In 100 mM NaCl, 50 mM Tris-HCl, 5 mM DTT, 4 mM reduced glutathione, 20% glycerol, pH 8.0.

Reconstitution

Following initial thaw, aliquot and freeze (-70°C).

Other Notes

Palka, H.L., et al. 2003. J. Biol. Chem.278, 5278.
Vadnais, J., et al. 2002. J. Biol. Chem.277, 48342.
Crostella, L., et al. 2001. Oncogene20, 3735.
Guiton, M., et al. 1994. J. Biol. Chem269, 30370.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service