Skip to Content
Merck
All Photos(5)

Key Documents

1.44158

Millipore

PyroDetect Interleukin Standard

Interleukin-1Beta standard for use in the Monocyte Activation Test (MAT) for Pyrogen testing

Synonym(s):

Interleukin-1β

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
€889.00

€889.00


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
1 EA
€889.00

About This Item

UNSPSC Code:
41116129
NACRES:
NA.24

€889.00


Estimated to ship on29 May 2025


Request more information

Quality Level

usage

sufficient for 20 tests

packaging

pkg of 1 vials

manufacturer/tradename

PyroDetect

application(s)

pharmaceutical
pyrogen testing

compatibility

configured for PyroDetect System

shipped in

ambient

storage temp.

2-8°C

suitability

pyrogenic bacteria

General description

PyroDetect Interleukin Standard is an Interleukin-1β Standard for control reactions in the Interleukin-1β ELISA. It is compatible for use with PyroDetect Kit. (921200), which is used for the detection and quantification of pyrogens based on the Monocyte-Activation Test (MAT).

Application

PyroDetect Interleukin Standard is used for Product Specific Validation with PyroDetect Kit, to check that the system actually detects interleukin-1β.

Storage and Stability

Store unopened vial at 2-8 °C. Do not use after expiration date of the product.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service