Skip to Content
Merck
All Photos(4)

Key Documents

1.44155

Millipore

PyroDetect Cryoblood

Cryopreserved blood pooled from 8 donors for Monocyte Activation Test (MAT), for Pyrogen testing, package of 2 vials

Synonym(s):

Cryopreserved blood

Sign Into View Organizational & Contract Pricing

Select a Size

2 X 2 ML
€295.00

€295.00


Estimated to ship on29 May 2025


Request more information

Select a Size

Change View
2 X 2 ML
€295.00

About This Item

UNSPSC Code:
41116129
NACRES:
NA.24

€295.00


Estimated to ship on29 May 2025


Request more information

Agency

EP Chapter 2.6.30

Quality Level

form

liquid

feature

ready-to-use

packaging

package of 2 vials
pkg of 2 vials

manufacturer/tradename

PyroDetect

application(s)

pharmaceutical
pyrogen testing

compatibility

configured for PyroDetect

shipped in

dry ice

storage temp.

-65 to -96°C

suitability

pyrogenic bacteria

General description

PyroDetect Cryoblood constitutes cryopreserved human whole blood pooled from 8 donors to detect pyrogens with the Monocyte-Activation Test (MAT). The cryopreserved and pooled human whole blood is tested for sterility and antigens of HIV, HAV, HCV, and HBV.

Application

The PyroDetect Cryoblood is part of the PyroDetect System and is used to detect pyrogens in pharmaceuticals, using lnterleukin-1β response.

Features and Benefits

Ready-to-use

Components

Cryoblood - deep frozen human blood preparation - 2 x 2 mL

Recommended products

PyroDetect Kit (1.44154)
Reference Standard Endotoxin (1.44161)
NEP Control of HKSA (MATHKSA)
NEP Control of Flagellin (MATFLAGELLIN)

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service